Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167127335-167127586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP27
Entrez Gene ID
92235
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
-
UniGene
Hs.632462
Gene Chromosome Location
Chr.1: 167094048-167129165 on Build GRCh38
Transcripts
NM_001080426.1 XM_005245603.3 XM_011510146.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5606494 c.2390C>T p.S797F Chr.1: 167127521-167127521 on Build GRCh38
6056680 c.2356C>G p.Q786E Chr.1: 167127487-167127487 on Build GRCh38
3863415 c.2318G>A p.G773E Chr.1: 167127449-167127449 on Build GRCh38
4796352 c.2291G>A p.G764E Chr.1: 167127422-167127422 on Build GRCh38
3477746 c.2293G>A p.D765N Chr.1: 167127424-167127424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769931637 Chr.1: 167127556-167127556 on Build GRCh38
rs761077185 Chr.1: 167127414-167127414 on Build GRCh38
rs938228337 Chr.1: 167127491-167127491 on Build GRCh38
rs148558531 Chr.1: 167127374-167127374 on Build GRCh38
rs139214807 Chr.1: 167127408-167127408 on Build GRCh38
rs201016877 Chr.1: 167127425-167127425 on Build GRCh38
rs747426096 Chr.1: 167127546-167127546 on Build GRCh38
rs762854287 Chr.1: 167127504-167127504 on Build GRCh38
rs766334312 Chr.1: 167127509-167127509 on Build GRCh38
rs771605750 Chr.1: 167127363-167127363 on Build GRCh38
rs140729613 Chr.1: 167127378-167127378 on Build GRCh38
rs752353847 Chr.1: 167127518-167127518 on Build GRCh38
rs916405075 Chr.1: 167127512-167127512 on Build GRCh38
rs374240399 Chr.1: 167127377-167127377 on Build GRCh38
rs199919968 Chr.1: 167127419-167127419 on Build GRCh38
rs768961009 Chr.1: 167127553-167127553 on Build GRCh38
rs775892985 Chr.1: 167127472-167127472 on Build GRCh38
rs142434634 Chr.1: 167127558-167127558 on Build GRCh38
rs747372678 Chr.1: 167127478-167127478 on Build GRCh38
rs981095950 Chr.1: 167127465-167127465 on Build GRCh38
rs761853640 Chr.1: 167127407-167127407 on Build GRCh38
rs755651147 Chr.1: 167127519-167127519 on Build GRCh38
rs146426286 Chr.1: 167127418-167127418 on Build GRCh38
rs918335205 Chr.1: 167127438-167127438 on Build GRCh38
rs758896566 Chr.1: 167127530-167127530 on Build GRCh38
rs148873336 Chr.1: 167127526-167127526 on Build GRCh38
rs145656152 Chr.1: 167127515-167127515 on Build GRCh38
rs773206628 Chr.1: 167127502-167127502 on Build GRCh38
rs772298564 Chr.1: 167127375-167127375 on Build GRCh38
rs79995360 Chr.1: 167127557-167127557 on Build GRCh38
rs200160038 Chr.1: 167127371-167127371 on Build GRCh38
rs751440284 Chr.1: 167127514-167127514 on Build GRCh38
rs747708985 Chr.1: 167127437-167127437 on Build GRCh38
rs145856876 Chr.1: 167127435-167127435 on Build GRCh38
rs779501819 Chr.1: 167127459-167127459 on Build GRCh38
rs765193414 Chr.1: 167127495-167127495 on Build GRCh38
rs201313034 Chr.1: 167127461-167127461 on Build GRCh38
rs201795536 Chr.1: 167127403-167127403 on Build GRCh38
rs538182309 Chr.1: 167127370-167127370 on Build GRCh38
rs200528135 Chr.1: 167127489-167127489 on Build GRCh38
rs755525549 Chr.1: 167127439-167127439 on Build GRCh38
rs754391838 Chr.1: 167127511-167127511 on Build GRCh38
rs768910128 Chr.1: 167127487-167127487 on Build GRCh38
rs772496141 Chr.1: 167127466-167127466 on Build GRCh38
rs754743221 Chr.1: 167127433-167127433 on Build GRCh38
rs781424729 Chr.1: 167127554-167127554 on Build GRCh38
rs371850525 Chr.1: 167127533-167127533 on Build GRCh38

View Full Product Details