Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203179354-203179587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHI3L1
Entrez Gene ID
1116
Gene Name
chitinase 3 like 1
Gene Aliases
ASRT7, CGP-39, GP-39, GP39, HC-gp39, HCGP-3P, YKL-40, YKL40, YYL-40, hCGP-39
UniGene
Hs.382202
Gene Chromosome Location
Chr.1: 203178931-203186794 on Build GRCh38
Transcripts
NM_001276.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5983823 c.1128C>T p.I376I Chr.1: 203179469-203179469 on Build GRCh38
4143245 c.1092T>C p.C364C Chr.1: 203179505-203179505 on Build GRCh38
1601539 c.1079A>T p.Q360L Chr.1: 203179518-203179518 on Build GRCh38
1601538 c.1091G>A p.C364Y Chr.1: 203179506-203179506 on Build GRCh38
902450 c.1098G>T p.Q366H Chr.1: 203179499-203179499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914557015 Chr.1: 203179410-203179410 on Build GRCh38
rs751034976 Chr.1: 203179448-203179448 on Build GRCh38
rs970312333 Chr.1: 203179373-203179373 on Build GRCh38
rs370960745 Chr.1: 203179515-203179515 on Build GRCh38
rs755036785 Chr.1: 203179475-203179475 on Build GRCh38
rs771141570 Chr.1: 203179478-203179478 on Build GRCh38
rs776842010 Chr.1: 203179381-203179381 on Build GRCh38
rs780043762 Chr.1: 203179464-203179464 on Build GRCh38
rs4950927 Chr.1: 203179505-203179505 on Build GRCh38
rs776309512 Chr.1: 203179453-203179453 on Build GRCh38
rs777003925 Chr.1: 203179409-203179409 on Build GRCh38
rs911106440 Chr.1: 203179437-203179437 on Build GRCh38
rs145648214 Chr.1: 203179453-203179453 on Build GRCh38
rs766276781 Chr.1: 203179514-203179514 on Build GRCh38
rs943974779 Chr.1: 203179438-203179438 on Build GRCh38
rs1003019596 Chr.1: 203179393-203179393 on Build GRCh38
rs756069288 Chr.1: 203179398-203179398 on Build GRCh38
rs763383345 Chr.1: 203179491-203179491 on Build GRCh38
rs148314652 Chr.1: 203179546-203179546 on Build GRCh38
rs888073208 Chr.1: 203179380-203179380 on Build GRCh38
rs763588973 Chr.1: 203179444-203179444 on Build GRCh38
rs774691241 Chr.1: 203179549-203179549 on Build GRCh38
rs778745033 Chr.1: 203179521-203179521 on Build GRCh38
rs768249525 Chr.1: 203179447-203179447 on Build GRCh38
rs771065423 Chr.1: 203179407-203179407 on Build GRCh38
rs138163817 Chr.1: 203179538-203179538 on Build GRCh38
rs1029881372 Chr.1: 203179446-203179446 on Build GRCh38
rs758194056 Chr.1: 203179421-203179429 on Build GRCh38
rs769168056 Chr.1: 203179548-203179548 on Build GRCh38
rs572689736 Chr.1: 203179457-203179457 on Build GRCh38
rs376352545 Chr.1: 203179554-203179554 on Build GRCh38
rs761616765 Chr.1: 203179502-203179502 on Build GRCh38
rs530402801 Chr.1: 203179527-203179527 on Build GRCh38
rs758355079 Chr.1: 203179505-203179507 on Build GRCh38
rs756333525 Chr.1: 203179540-203179540 on Build GRCh38
rs781387316 Chr.1: 203179529-203179529 on Build GRCh38
rs201313181 Chr.1: 203179492-203179492 on Build GRCh38
rs1048181516 Chr.1: 203179470-203179470 on Build GRCh38
rs112567571 Chr.1: 203179552-203179552 on Build GRCh38
rs747254602 Chr.1: 203179405-203179405 on Build GRCh38
rs745661413 Chr.1: 203179394-203179394 on Build GRCh38
rs761261967 Chr.1: 203179493-203179493 on Build GRCh38
rs146151706 Chr.1: 203179553-203179553 on Build GRCh38
rs752716990 Chr.1: 203179523-203179523 on Build GRCh38
rs767096746 Chr.1: 203179504-203179504 on Build GRCh38
rs947717704 Chr.1: 203179503-203179503 on Build GRCh38
rs755942899 Chr.1: 203179462-203179462 on Build GRCh38
rs780027741 Chr.1: 203179435-203179445 on Build GRCh38
rs202080664 Chr.1: 203179422-203179422 on Build GRCh38
rs749279657 Chr.1: 203179400-203179400 on Build GRCh38
rs753818436 Chr.1: 203179469-203179469 on Build GRCh38
rs746146575 Chr.1: 203179485-203179485 on Build GRCh38
rs754908889 Chr.1: 203179517-203179517 on Build GRCh38
rs761469662 Chr.1: 203179449-203179449 on Build GRCh38
rs377080511 Chr.1: 203179399-203179399 on Build GRCh38
rs563362796 Chr.1: 203179544-203179544 on Build GRCh38
rs200560076 Chr.1: 203179423-203179423 on Build GRCh38
rs968742730 Chr.1: 203179466-203179466 on Build GRCh38
rs770126580 Chr.1: 203179487-203179487 on Build GRCh38
rs377668825 Chr.1: 203179476-203179476 on Build GRCh38
rs746966964 Chr.1: 203179438-203179438 on Build GRCh38
rs1049410 Chr.1: 203179556-203179556 on Build GRCh38
rs781236852 Chr.1: 203179482-203179482 on Build GRCh38
rs370300790 Chr.1: 203179404-203179404 on Build GRCh38
rs527531482 Chr.1: 203179557-203179557 on Build GRCh38
rs747751870 Chr.1: 203179455-203179459 on Build GRCh38
rs376775749 Chr.1: 203179430-203179430 on Build GRCh38

View Full Product Details