Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 82138473-82138746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFL1
Entrez Gene ID
79631
Gene Name
elongation factor like GTPase 1
Gene Aliases
EFTUD1, FAM42A, HsT19294, RIA1
UniGene
Hs.459114
Gene Chromosome Location
Chr.15: 82130220-82262763 on Build GRCh38
Transcripts
NM_001040610.2 NM_001322844.1 NM_001322845.1 NM_024580.5 NR_136410.1 XM_011522000.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
701619 c.3142G>C p.A1048P Chr.15: 82138690-82138690 on Build GRCh38
2012176 c.3139C>A p.L1047M Chr.15: 82138693-82138693 on Build GRCh38
398942 c.3115G>A p.E1039K Chr.15: 82138717-82138717 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs183657692 Chr.15: 82138549-82138549 on Build GRCh38
rs116987284 Chr.15: 82138621-82138621 on Build GRCh38
rs964327938 Chr.15: 82138602-82138602 on Build GRCh38
rs200449747 Chr.15: 82138668-82138668 on Build GRCh38
rs750079244 Chr.15: 82138658-82138658 on Build GRCh38
rs762768501 Chr.15: 82138628-82138628 on Build GRCh38
rs780164107 Chr.15: 82138540-82138540 on Build GRCh38
rs539508832 Chr.15: 82138536-82138536 on Build GRCh38
rs749634842 Chr.15: 82138666-82138666 on Build GRCh38
rs772190046 Chr.15: 82138693-82138693 on Build GRCh38
rs780538685 Chr.15: 82138664-82138664 on Build GRCh38
rs139573495 Chr.15: 82138530-82138530 on Build GRCh38
rs779505479 Chr.15: 82138679-82138679 on Build GRCh38
rs775303934 Chr.15: 82138622-82138622 on Build GRCh38
rs201517223 Chr.15: 82138715-82138715 on Build GRCh38
rs573035571 Chr.15: 82138609-82138609 on Build GRCh38
rs763967001 Chr.15: 82138631-82138631 on Build GRCh38
rs773212031 Chr.15: 82138703-82138703 on Build GRCh38
rs774626448 Chr.15: 82138639-82138639 on Build GRCh38
rs1019880171 Chr.15: 82138566-82138566 on Build GRCh38
rs111858844 Chr.15: 82138638-82138638 on Build GRCh38
rs919176906 Chr.15: 82138525-82138525 on Build GRCh38
rs767001143 Chr.15: 82138656-82138656 on Build GRCh38
rs756717349 Chr.15: 82138649-82138649 on Build GRCh38
rs748608478 Chr.15: 82138688-82138688 on Build GRCh38
rs975988293 Chr.15: 82138515-82138515 on Build GRCh38
rs997009368 Chr.15: 82138617-82138617 on Build GRCh38
rs938856997 Chr.15: 82138672-82138672 on Build GRCh38
rs967074842 Chr.15: 82138699-82138699 on Build GRCh38

View Full Product Details