Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 30560002-30560227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf21
Entrez Gene ID
80231
Gene Name
chromosome X open reading frame 21
Gene Aliases
-
UniGene
Hs.665009
Gene Chromosome Location
Chr.X: 30558824-30577916 on Build GRCh38
Transcripts
NM_025159.2

SNP Detail

SNP ID Chromosome Location
rs140764539 Chr.X: 30560111-30560111 on Build GRCh38
rs770745577 Chr.X: 30560201-30560201 on Build GRCh38
rs770694258 Chr.X: 30560042-30560042 on Build GRCh38
rs748604114 Chr.X: 30560164-30560164 on Build GRCh38
rs950393150 Chr.X: 30560070-30560070 on Build GRCh38
rs935933392 Chr.X: 30560170-30560170 on Build GRCh38
rs748637871 Chr.X: 30560027-30560027 on Build GRCh38
rs758681466 Chr.X: 30560110-30560110 on Build GRCh38
rs1053468612 Chr.X: 30560173-30560173 on Build GRCh38
rs774070004 Chr.X: 30560062-30560062 on Build GRCh38
rs373780978 Chr.X: 30560137-30560137 on Build GRCh38
rs751682389 Chr.X: 30560115-30560115 on Build GRCh38
rs138820651 Chr.X: 30560191-30560191 on Build GRCh38
rs752601692 Chr.X: 30560084-30560084 on Build GRCh38
rs770082070 Chr.X: 30560030-30560030 on Build GRCh38
rs762706999 Chr.X: 30560036-30560036 on Build GRCh38
rs768160860 Chr.X: 30560190-30560190 on Build GRCh38
rs753576613 Chr.X: 30560108-30560108 on Build GRCh38
rs150122685 Chr.X: 30560133-30560133 on Build GRCh38
rs767540673 Chr.X: 30560082-30560082 on Build GRCh38
rs758937552 Chr.X: 30560067-30560067 on Build GRCh38
rs892451272 Chr.X: 30560182-30560182 on Build GRCh38
rs773641205 Chr.X: 30560033-30560033 on Build GRCh38
rs756582268 Chr.X: 30560171-30560171 on Build GRCh38

View Full Product Details