Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3814257-3814530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCBP3
Entrez Gene ID
55421
Gene Name
nuclear cap binding subunit 3
Gene Aliases
C17orf85, ELG, HSA277841
UniGene
Hs.120963
Gene Chromosome Location
Chr.17: 3806748-3846251 on Build GRCh38
Transcripts
NM_001114118.2 XM_005256717.2 XM_006721556.2 XM_011523954.2 XM_011523955.2 XM_017024826.1 XM_017024827.1 XM_017024828.1 XM_017024829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198273 c.631C>T p.R211C Chr.17: 3814478-3814478 on Build GRCh38
978522 c.691G>A p.V231I Chr.17: 3814418-3814418 on Build GRCh38
6614773 c.1547C>T p.S516F Chr.17: 3814402-3814402 on Build GRCh38
1198275 c.650G>T p.R217I Chr.17: 3814459-3814459 on Build GRCh38
1588807 c.1472G>A p.R491H Chr.17: 3814477-3814477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769525305 Chr.17: 3814302-3814302 on Build GRCh38
rs750842675 Chr.17: 3814437-3814437 on Build GRCh38
rs750710112 Chr.17: 3814447-3814447 on Build GRCh38
rs747260140 Chr.17: 3814292-3814292 on Build GRCh38
rs764499530 Chr.17: 3814345-3814345 on Build GRCh38
rs755178793 Chr.17: 3814467-3814467 on Build GRCh38
rs569227083 Chr.17: 3814346-3814346 on Build GRCh38
rs1022453085 Chr.17: 3814495-3814495 on Build GRCh38
rs773895980 Chr.17: 3814333-3814333 on Build GRCh38
rs760341775 Chr.17: 3814415-3814415 on Build GRCh38
rs780041254 Chr.17: 3814446-3814446 on Build GRCh38
rs1043946515 Chr.17: 3814353-3814353 on Build GRCh38
rs770593185 Chr.17: 3814501-3814501 on Build GRCh38
rs147672066 Chr.17: 3814422-3814422 on Build GRCh38
rs758366075 Chr.17: 3814373-3814373 on Build GRCh38
rs781440938 Chr.17: 3814480-3814480 on Build GRCh38
rs372409573 Chr.17: 3814331-3814331 on Build GRCh38
rs1011246746 Chr.17: 3814494-3814494 on Build GRCh38
rs374828748 Chr.17: 3814380-3814380 on Build GRCh38
rs757781733 Chr.17: 3814347-3814347 on Build GRCh38
rs773004471 Chr.17: 3814303-3814303 on Build GRCh38
rs768869229 Chr.17: 3814387-3814387 on Build GRCh38
rs915371021 Chr.17: 3814280-3814280 on Build GRCh38
rs749080595 Chr.17: 3814499-3814499 on Build GRCh38
rs779336127 Chr.17: 3814350-3814350 on Build GRCh38
rs938263441 Chr.17: 3814376-3814376 on Build GRCh38
rs749093702 Chr.17: 3814396-3814396 on Build GRCh38
rs914055367 Chr.17: 3814343-3814343 on Build GRCh38
rs766292607 Chr.17: 3814332-3814332 on Build GRCh38
rs780126300 Chr.17: 3814378-3814378 on Build GRCh38
rs184645400 Chr.17: 3814344-3814344 on Build GRCh38
rs747176507 Chr.17: 3814379-3814379 on Build GRCh38
rs758953634 Chr.17: 3814442-3814442 on Build GRCh38
rs139309920 Chr.17: 3814390-3814390 on Build GRCh38
rs532604306 Chr.17: 3814337-3814337 on Build GRCh38
rs566972085 Chr.17: 3814336-3814336 on Build GRCh38
rs774197585 Chr.17: 3814404-3814404 on Build GRCh38
rs745735124 Chr.17: 3814508-3814508 on Build GRCh38
rs901533761 Chr.17: 3814430-3814430 on Build GRCh38
rs557284004 Chr.17: 3814402-3814402 on Build GRCh38
rs149973573 Chr.17: 3814413-3814413 on Build GRCh38
rs765662426 Chr.17: 3814425-3814425 on Build GRCh38
rs762809591 Chr.17: 3814304-3814304 on Build GRCh38
rs765334742 Chr.17: 3814424-3814424 on Build GRCh38
rs775157987 Chr.17: 3814414-3814414 on Build GRCh38
rs755849415 Chr.17: 3814309-3814309 on Build GRCh38
rs1016108038 Chr.17: 3814502-3814502 on Build GRCh38
rs370718531 Chr.17: 3814506-3814506 on Build GRCh38
rs763903447 Chr.17: 3814354-3814354 on Build GRCh38
rs745927502 Chr.17: 3814361-3814361 on Build GRCh38
rs763908946 Chr.17: 3814418-3814418 on Build GRCh38
rs184111975 Chr.17: 3814477-3814477 on Build GRCh38
rs753731334 Chr.17: 3814487-3814488 on Build GRCh38
rs754244716 Chr.17: 3814419-3814419 on Build GRCh38

View Full Product Details