Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104897282-104897513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP12
Entrez Gene ID
100506742
Gene Name
caspase 12 (gene/pseudogene)
Gene Aliases
CASP-12, CASP12P1
UniGene
Hs.476989
Gene Chromosome Location
Chr.11: 104885718-104898670 on Build GRCh38
Transcripts
NM_001191016.2 NR_034061.2 NR_034063.2 NR_034064.2 NR_034065.2 NR_034066.2 NR_034067.2 NR_034068.2 NR_034070.2 NR_034071.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4597624 c.62C>T p.T21I Chr.11: 104897414-104897414 on Build GRCh38
6519534 c.60G>T p.K20N Chr.11: 104897416-104897416 on Build GRCh38
3375622 c.35T>A p.V12D Chr.11: 104897441-104897441 on Build GRCh38
5461908 c.28G>A p.V10I Chr.11: 104897448-104897448 on Build GRCh38
2109059 c.29T>C p.V10A Chr.11: 104897447-104897447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980048404 Chr.11: 104897435-104897435 on Build GRCh38
rs758171254 Chr.11: 104897409-104897410 on Build GRCh38
rs1015057842 Chr.11: 104897369-104897369 on Build GRCh38
rs932970140 Chr.11: 104897313-104897313 on Build GRCh38
rs768226488 Chr.11: 104897416-104897416 on Build GRCh38
rs1022930071 Chr.11: 104897434-104897434 on Build GRCh38
rs780517972 Chr.11: 104897322-104897322 on Build GRCh38
rs574759342 Chr.11: 104897454-104897454 on Build GRCh38
rs769378428 Chr.11: 104897373-104897373 on Build GRCh38
rs905884810 Chr.11: 104897436-104897436 on Build GRCh38
rs115063404 Chr.11: 104897378-104897378 on Build GRCh38
rs758946913 Chr.11: 104897318-104897318 on Build GRCh38
rs138698464 Chr.11: 104897414-104897414 on Build GRCh38
rs17103291 Chr.11: 104897392-104897392 on Build GRCh38
rs555367 Chr.11: 104897371-104897371 on Build GRCh38
rs267602676 Chr.11: 104897431-104897431 on Build GRCh38
rs761071405 Chr.11: 104897425-104897425 on Build GRCh38
rs142724970 Chr.11: 104897427-104897427 on Build GRCh38
rs989850622 Chr.11: 104897452-104897452 on Build GRCh38
rs926913285 Chr.11: 104897445-104897445 on Build GRCh38
rs114027557 Chr.11: 104897384-104897384 on Build GRCh38
rs987191544 Chr.11: 104897332-104897332 on Build GRCh38
rs910219104 Chr.11: 104897334-104897334 on Build GRCh38
rs775587686 Chr.11: 104897451-104897451 on Build GRCh38
rs774352791 Chr.11: 104897413-104897413 on Build GRCh38

View Full Product Details