Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 199924962-199925233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf69
Entrez Gene ID
205327
Gene Name
chromosome 2 open reading frame 69
Gene Aliases
-
UniGene
Hs.154494
Gene Chromosome Location
Chr.2: 199911256-199928273 on Build GRCh38
Transcripts
NM_153689.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
718599 c.387G>T p.W129C Chr.2: 199925115-199925115 on Build GRCh38
296275 c.356G>A p.R119H Chr.2: 199925084-199925084 on Build GRCh38
4583076 c.488A>G p.Y163C Chr.2: 199925216-199925216 on Build GRCh38
4090047 c.355C>T p.R119C Chr.2: 199925083-199925083 on Build GRCh38
6409863 c.356G>C p.R119P Chr.2: 199925084-199925084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775537788 Chr.2: 199925166-199925166 on Build GRCh38
rs746476821 Chr.2: 199925136-199925136 on Build GRCh38
rs758028301 Chr.2: 199925132-199925132 on Build GRCh38
rs770710492 Chr.2: 199925137-199925137 on Build GRCh38
rs764130811 Chr.2: 199925061-199925061 on Build GRCh38
rs371259262 Chr.2: 199925051-199925051 on Build GRCh38
rs148073145 Chr.2: 199925081-199925081 on Build GRCh38
rs200237620 Chr.2: 199925026-199925026 on Build GRCh38
rs190789042 Chr.2: 199925103-199925103 on Build GRCh38
rs762047776 Chr.2: 199925083-199925083 on Build GRCh38
rs900737861 Chr.2: 199925052-199925052 on Build GRCh38
rs762856792 Chr.2: 199925168-199925168 on Build GRCh38
rs200098289 Chr.2: 199925135-199925135 on Build GRCh38
rs768533781 Chr.2: 199925189-199925189 on Build GRCh38
rs759186973 Chr.2: 199925039-199925039 on Build GRCh38
rs186306624 Chr.2: 199925158-199925158 on Build GRCh38
rs769473017 Chr.2: 199925159-199925159 on Build GRCh38
rs780674554 Chr.2: 199925111-199925111 on Build GRCh38
rs763159764 Chr.2: 199925058-199925058 on Build GRCh38
rs187875052 Chr.2: 199925089-199925089 on Build GRCh38
rs754264597 Chr.2: 199925119-199925119 on Build GRCh38
rs767688764 Chr.2: 199925084-199925084 on Build GRCh38
rs780651524 Chr.2: 199925150-199925150 on Build GRCh38
rs377702460 Chr.2: 199925044-199925044 on Build GRCh38
rs534813908 Chr.2: 199925014-199925014 on Build GRCh38
rs905441673 Chr.2: 199925109-199925109 on Build GRCh38

View Full Product Details