Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30922887-30923126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAP2
Entrez Gene ID
55803
Gene Name
ArfGAP with dual PH domains 2
Gene Aliases
CENTA2, HSA272195, cent-b
UniGene
Hs.514063
Gene Chromosome Location
Chr.17: 30921736-30959193 on Build GRCh38
Transcripts
NM_018404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3396262 c.152T>A p.V51D Chr.17: 30922997-30922997 on Build GRCh38
7127539 c.223G>C p.E75Q Chr.17: 30923068-30923068 on Build GRCh38
4579571 c.225+1G>A p.? Chr.17: 30923071-30923071 on Build GRCh38
4839380 c.200A>T p.D67V Chr.17: 30923045-30923045 on Build GRCh38
6594617 c.199G>T p.D67Y Chr.17: 30923044-30923044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535948357 Chr.17: 30922991-30922991 on Build GRCh38
rs2232271 Chr.17: 30923083-30923083 on Build GRCh38
rs201030071 Chr.17: 30923087-30923087 on Build GRCh38
rs200602165 Chr.17: 30923084-30923084 on Build GRCh38
rs997032195 Chr.17: 30923039-30923039 on Build GRCh38
rs1019426967 Chr.17: 30922985-30922985 on Build GRCh38
rs778958039 Chr.17: 30922982-30922982 on Build GRCh38
rs772979991 Chr.17: 30923038-30923038 on Build GRCh38
rs779784183 Chr.17: 30922968-30922968 on Build GRCh38
rs148209227 Chr.17: 30923026-30923026 on Build GRCh38
rs1039075985 Chr.17: 30922993-30922993 on Build GRCh38
rs370692296 Chr.17: 30922917-30922917 on Build GRCh38
rs765197388 Chr.17: 30923063-30923063 on Build GRCh38
rs766505175 Chr.17: 30922908-30922908 on Build GRCh38
rs963813526 Chr.17: 30922989-30922989 on Build GRCh38
rs753100365 Chr.17: 30922942-30922942 on Build GRCh38
rs143073788 Chr.17: 30922945-30922945 on Build GRCh38
rs746751935 Chr.17: 30923048-30923048 on Build GRCh38
rs1021287166 Chr.17: 30923043-30923043 on Build GRCh38
rs201918740 Chr.17: 30923098-30923098 on Build GRCh38
rs751975930 Chr.17: 30923079-30923079 on Build GRCh38
rs771923291 Chr.17: 30923033-30923033 on Build GRCh38
rs901595228 Chr.17: 30923023-30923023 on Build GRCh38
rs754397315 Chr.17: 30922977-30922977 on Build GRCh38
rs776681714 Chr.17: 30923055-30923055 on Build GRCh38
rs142699740 Chr.17: 30923069-30923069 on Build GRCh38
rs374371980 Chr.17: 30923107-30923107 on Build GRCh38
rs765809462 Chr.17: 30922935-30922935 on Build GRCh38
rs753409348 Chr.17: 30923085-30923085 on Build GRCh38
rs199959488 Chr.17: 30923086-30923086 on Build GRCh38
rs150091453 Chr.17: 30922951-30922951 on Build GRCh38
rs375536248 Chr.17: 30922936-30922936 on Build GRCh38
rs748858882 Chr.17: 30922975-30922975 on Build GRCh38
rs759687818 Chr.17: 30923056-30923056 on Build GRCh38
rs759469734 Chr.17: 30922926-30922926 on Build GRCh38
rs566058284 Chr.17: 30923073-30923073 on Build GRCh38
rs146923575 Chr.17: 30923071-30923071 on Build GRCh38
rs750156060 Chr.17: 30922953-30922953 on Build GRCh38

View Full Product Details