Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31116005-31116258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDSN
Entrez Gene ID
1041
Gene Name
corneodesmosin
Gene Aliases
HTSS, HTSS1, HYPT2, PSS, PSS1, S
UniGene
Hs.310958
Gene Chromosome Location
Chr.6: 31115088-31120475 on Build GRCh38
Transcripts
NM_001264.4 NM_014068.2

Gene Symbol
PSORS1C1
Entrez Gene ID
170679
Gene Name
psoriasis susceptibility 1 candidate 1
Gene Aliases
C6orf16, SEEK1
UniGene
Hs.310958
Gene Chromosome Location
Chr.6: 31114831-31140092 on Build GRCh38
Transcripts
NM_001264.4 NM_014068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1443215 c.1473T>C p.A491A Chr.6: 31116142-31116142 on Build GRCh38
3928319 c.1440C>T p.S480S Chr.6: 31116175-31116175 on Build GRCh38
5696254 c.1558C>T p.L520L Chr.6: 31116057-31116057 on Build GRCh38
6359571 c.1365T>C p.S455S Chr.6: 31116250-31116250 on Build GRCh38
3252658 c.1490G>A p.R497H Chr.6: 31116125-31116125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760397250 Chr.6: 31116063-31116063 on Build GRCh38
rs779015672 Chr.6: 31116042-31116042 on Build GRCh38
rs565620030 Chr.6: 31116047-31116047 on Build GRCh38
rs150111690 Chr.6: 31116100-31116100 on Build GRCh38
rs985939032 Chr.6: 31116104-31116104 on Build GRCh38
rs748019758 Chr.6: 31116131-31116131 on Build GRCh38
rs1012807064 Chr.6: 31116227-31116227 on Build GRCh38
rs753114787 Chr.6: 31116093-31116093 on Build GRCh38
rs200654068 Chr.6: 31116126-31116126 on Build GRCh38
rs774392868 Chr.6: 31116128-31116128 on Build GRCh38
rs199905483 Chr.6: 31116202-31116202 on Build GRCh38
rs145583110 Chr.6: 31116168-31116168 on Build GRCh38
rs766061104 Chr.6: 31116064-31116064 on Build GRCh38
rs141505496 Chr.6: 31116151-31116151 on Build GRCh38
rs781428949 Chr.6: 31116234-31116234 on Build GRCh38
rs868580577 Chr.6: 31116084-31116084 on Build GRCh38
rs918579308 Chr.6: 31116198-31116198 on Build GRCh38
rs750279345 Chr.6: 31116035-31116035 on Build GRCh38
rs761422763 Chr.6: 31116050-31116050 on Build GRCh38
rs370187632 Chr.6: 31116182-31116182 on Build GRCh38
rs138093208 Chr.6: 31116174-31116174 on Build GRCh38
rs760204848 Chr.6: 31116163-31116163 on Build GRCh38
rs772973219 Chr.6: 31116055-31116055 on Build GRCh38
rs753307421 Chr.6: 31116186-31116186 on Build GRCh38
rs892022806 Chr.6: 31116120-31116120 on Build GRCh38
rs780284826 Chr.6: 31116117-31116129 on Build GRCh38
rs1043716279 Chr.6: 31116171-31116171 on Build GRCh38
rs750619040 Chr.6: 31116236-31116236 on Build GRCh38
rs768301621 Chr.6: 31116125-31116125 on Build GRCh38
rs755064204 Chr.6: 31116197-31116197 on Build GRCh38
rs896582108 Chr.6: 31116209-31116209 on Build GRCh38
rs751655884 Chr.6: 31116206-31116206 on Build GRCh38
rs142570009 Chr.6: 31116203-31116203 on Build GRCh38
rs756296748 Chr.6: 31116085-31116085 on Build GRCh38
rs775263614 Chr.6: 31116030-31116030 on Build GRCh38
rs150958659 Chr.6: 31116090-31116090 on Build GRCh38
rs757281835 Chr.6: 31116212-31116212 on Build GRCh38
rs771677105 Chr.6: 31116145-31116145 on Build GRCh38
rs753865872 Chr.6: 31116121-31116121 on Build GRCh38
rs772851382 Chr.6: 31116160-31116160 on Build GRCh38
rs765498828 Chr.6: 31116185-31116185 on Build GRCh38
rs138408228 Chr.6: 31116117-31116117 on Build GRCh38
rs768599029 Chr.6: 31116040-31116040 on Build GRCh38
rs754267246 Chr.6: 31116083-31116083 on Build GRCh38
rs942054023 Chr.6: 31116082-31116082 on Build GRCh38
rs375368228 Chr.6: 31116089-31116089 on Build GRCh38
rs766911427 Chr.6: 31116052-31116052 on Build GRCh38
rs757477487 Chr.6: 31116116-31116116 on Build GRCh38
rs909107761 Chr.6: 31116148-31116148 on Build GRCh38
rs140801967 Chr.6: 31116099-31116099 on Build GRCh38
rs3130981 Chr.6: 31116036-31116036 on Build GRCh38
rs976237458 Chr.6: 31116092-31116092 on Build GRCh38

View Full Product Details