Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53292003-53292276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP1A
Entrez Gene ID
55802
Gene Name
decapping mRNA 1A
Gene Aliases
HSA275986, Nbla00360, SMAD4IP1, SMIF
UniGene
Hs.476353
Gene Chromosome Location
Chr.3: 53283423-53347627 on Build GRCh38
Transcripts
NM_001290204.1 NM_001290205.1 NM_001290206.1 NM_001290207.1 NM_018403.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640964 c.1270G>A p.A424T Chr.3: 53292182-53292182 on Build GRCh38
1047277 c.1268C>T p.P423L Chr.3: 53292184-53292184 on Build GRCh38
337861 c.1233A>G p.P411P Chr.3: 53292219-53292219 on Build GRCh38
6544481 c.1328C>A p.A443E Chr.3: 53292124-53292124 on Build GRCh38
1194101 c.1325C>G p.A442G Chr.3: 53292127-53292127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376012299 Chr.3: 53292084-53292084 on Build GRCh38
rs781980864 Chr.3: 53292138-53292138 on Build GRCh38
rs782087639 Chr.3: 53292144-53292144 on Build GRCh38
rs375539952 Chr.3: 53292241-53292241 on Build GRCh38
rs368534716 Chr.3: 53292204-53292204 on Build GRCh38
rs191456151 Chr.3: 53292047-53292047 on Build GRCh38
rs782457634 Chr.3: 53292186-53292186 on Build GRCh38
rs781823778 Chr.3: 53292184-53292184 on Build GRCh38
rs781928933 Chr.3: 53292149-53292149 on Build GRCh38
rs74920529 Chr.3: 53292190-53292190 on Build GRCh38
rs782139654 Chr.3: 53292221-53292221 on Build GRCh38
rs782397219 Chr.3: 53292203-53292203 on Build GRCh38
rs782012601 Chr.3: 53292045-53292045 on Build GRCh38
rs375532510 Chr.3: 53292187-53292187 on Build GRCh38
rs782046290 Chr.3: 53292039-53292039 on Build GRCh38
rs782533345 Chr.3: 53292201-53292201 on Build GRCh38
rs370368782 Chr.3: 53292095-53292095 on Build GRCh38
rs181600590 Chr.3: 53292054-53292054 on Build GRCh38
rs782352848 Chr.3: 53292206-53292206 on Build GRCh38
rs782664949 Chr.3: 53292083-53292083 on Build GRCh38
rs554234163 Chr.3: 53292110-53292110 on Build GRCh38
rs781972678 Chr.3: 53292231-53292231 on Build GRCh38
rs35623983 Chr.3: 53292034-53292035 on Build GRCh38
rs186298829 Chr.3: 53292070-53292070 on Build GRCh38
rs781987714 Chr.3: 53292044-53292044 on Build GRCh38
rs746165974 Chr.3: 53292117-53292117 on Build GRCh38
rs782383240 Chr.3: 53292125-53292125 on Build GRCh38
rs782585435 Chr.3: 53292209-53292209 on Build GRCh38
rs782741683 Chr.3: 53292183-53292183 on Build GRCh38
rs543268570 Chr.3: 53292165-53292165 on Build GRCh38
rs371233245 Chr.3: 53292175-53292175 on Build GRCh38
rs782817511 Chr.3: 53292043-53292043 on Build GRCh38
rs373828434 Chr.3: 53292106-53292106 on Build GRCh38
rs782027053 Chr.3: 53292219-53292219 on Build GRCh38
rs782462657 Chr.3: 53292062-53292062 on Build GRCh38
rs782724752 Chr.3: 53292146-53292146 on Build GRCh38
rs369792629 Chr.3: 53292065-53292065 on Build GRCh38
rs782125157 Chr.3: 53292049-53292049 on Build GRCh38
rs80051098 Chr.3: 53292191-53292191 on Build GRCh38
rs782533530 Chr.3: 53292079-53292079 on Build GRCh38
rs781835605 Chr.3: 53292057-53292057 on Build GRCh38
rs782233691 Chr.3: 53292035-53292035 on Build GRCh38
rs781983049 Chr.3: 53292214-53292214 on Build GRCh38
rs781936832 Chr.3: 53292037-53292037 on Build GRCh38
rs781910772 Chr.3: 53292077-53292077 on Build GRCh38
rs782083952 Chr.3: 53292233-53292233 on Build GRCh38
rs782039030 Chr.3: 53292155-53292155 on Build GRCh38
rs782602453 Chr.3: 53292099-53292099 on Build GRCh38
rs782270207 Chr.3: 53292115-53292115 on Build GRCh38
rs143190624 Chr.3: 53292112-53292112 on Build GRCh38
rs782347448 Chr.3: 53292036-53292036 on Build GRCh38
rs76231067 Chr.3: 53292236-53292236 on Build GRCh38
rs781875404 Chr.3: 53292170-53292170 on Build GRCh38
rs781902746 Chr.3: 53292197-53292197 on Build GRCh38
rs782380035 Chr.3: 53292225-53292225 on Build GRCh38
rs782311069 Chr.3: 53292111-53292111 on Build GRCh38
rs371247382 Chr.3: 53292237-53292237 on Build GRCh38

View Full Product Details