Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3492189-3492428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOHH
Entrez Gene ID
83475
Gene Name
deoxyhypusine hydroxylase/monooxygenase
Gene Aliases
HLRC1, hDOHH
UniGene
Hs.515064
Gene Chromosome Location
Chr.19: 3490821-3500940 on Build GRCh38
Transcripts
NM_001145165.1 NM_031304.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6375035 c.550A>G p.N184D Chr.19: 3492301-3492301 on Build GRCh38
1612046 c.504G>T p.E168D Chr.19: 3492347-3492347 on Build GRCh38
6440998 c.458C>T p.A153V Chr.19: 3492393-3492393 on Build GRCh38
1204426 c.452C>T p.P151L Chr.19: 3492399-3492399 on Build GRCh38
6201266 c.508C>T p.R170W Chr.19: 3492343-3492343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746100860 Chr.19: 3492335-3492335 on Build GRCh38
rs554577564 Chr.19: 3492370-3492370 on Build GRCh38
rs553950608 Chr.19: 3492396-3492396 on Build GRCh38
rs570290335 Chr.19: 3492331-3492331 on Build GRCh38
rs903836201 Chr.19: 3492360-3492360 on Build GRCh38
rs770738930 Chr.19: 3492322-3492322 on Build GRCh38
rs752135041 Chr.19: 3492385-3492385 on Build GRCh38
rs777073505 Chr.19: 3492346-3492346 on Build GRCh38
rs764571059 Chr.19: 3492263-3492263 on Build GRCh38
rs78287632 Chr.19: 3492320-3492320 on Build GRCh38
rs780673069 Chr.19: 3492298-3492298 on Build GRCh38
rs778757736 Chr.19: 3492405-3492405 on Build GRCh38
rs772339647 Chr.19: 3492336-3492336 on Build GRCh38
rs767674710 Chr.19: 3492230-3492230 on Build GRCh38
rs758104640 Chr.19: 3492281-3492281 on Build GRCh38
rs78690374 Chr.19: 3492245-3492245 on Build GRCh38
rs561886243 Chr.19: 3492380-3492380 on Build GRCh38
rs1052946856 Chr.19: 3492324-3492324 on Build GRCh38
rs777547733 Chr.19: 3492313-3492313 on Build GRCh38
rs140350778 Chr.19: 3492256-3492257 on Build GRCh38
rs202060994 Chr.19: 3492221-3492221 on Build GRCh38
rs753777762 Chr.19: 3492398-3492398 on Build GRCh38
rs1002221408 Chr.19: 3492366-3492366 on Build GRCh38
rs576963088 Chr.19: 3492401-3492401 on Build GRCh38
rs748141704 Chr.19: 3492299-3492299 on Build GRCh38
rs1051808 Chr.19: 3492342-3492342 on Build GRCh38
rs751200106 Chr.19: 3492290-3492290 on Build GRCh38
rs533833325 Chr.19: 3492379-3492379 on Build GRCh38
rs769250530 Chr.19: 3492343-3492343 on Build GRCh38
rs775656921 Chr.19: 3492338-3492338 on Build GRCh38
rs111669819 Chr.19: 3492339-3492339 on Build GRCh38
rs765896061 Chr.19: 3492289-3492289 on Build GRCh38
rs919477799 Chr.19: 3492215-3492215 on Build GRCh38
rs1034143712 Chr.19: 3492373-3492373 on Build GRCh38
rs755986071 Chr.19: 3492310-3492310 on Build GRCh38
rs908864853 Chr.19: 3492254-3492254 on Build GRCh38
rs754519502 Chr.19: 3492297-3492297 on Build GRCh38
rs965500521 Chr.19: 3492384-3492384 on Build GRCh38
rs754086898 Chr.19: 3492279-3492279 on Build GRCh38
rs867679784 Chr.19: 3492232-3492232 on Build GRCh38
rs373704209 Chr.19: 3492374-3492374 on Build GRCh38
rs756076115 Chr.19: 3492395-3492395 on Build GRCh38
rs761254210 Chr.19: 3492259-3492259 on Build GRCh38
rs757068576 Chr.19: 3492402-3492402 on Build GRCh38
rs536165069 Chr.19: 3492349-3492349 on Build GRCh38
rs1039166226 Chr.19: 3492287-3492287 on Build GRCh38
rs943078444 Chr.19: 3492272-3492272 on Build GRCh38

View Full Product Details