Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 673216-673487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5I
Entrez Gene ID
521
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit E
Gene Aliases
ATP5K
UniGene
Hs.85539
Gene Chromosome Location
Chr.4: 672436-674338 on Build GRCh38
Transcripts
NM_007100.3 NR_033743.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
138420 c.187G>A p.E63K Chr.4: 673306-673306 on Build GRCh38
5892947 c.118G>A p.E40K Chr.4: 673375-673375 on Build GRCh38
4664430 c.178G>T p.E60* Chr.4: 673315-673315 on Build GRCh38
1056665 c.119A>G p.E40G Chr.4: 673374-673374 on Build GRCh38
1056664 c.137A>G p.E46G Chr.4: 673356-673356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762507288 Chr.4: 673341-673341 on Build GRCh38
rs200608822 Chr.4: 673384-673384 on Build GRCh38
rs370751906 Chr.4: 673325-673325 on Build GRCh38
rs753138965 Chr.4: 673284-673284 on Build GRCh38
rs562074822 Chr.4: 673445-673445 on Build GRCh38
rs767660093 Chr.4: 673279-673280 on Build GRCh38
rs780085696 Chr.4: 673376-673376 on Build GRCh38
rs776161747 Chr.4: 673370-673370 on Build GRCh38
rs139451905 Chr.4: 673242-673242 on Build GRCh38
rs756511791 Chr.4: 673286-673286 on Build GRCh38
rs1047586669 Chr.4: 673415-673415 on Build GRCh38
rs113258489 Chr.4: 673300-673300 on Build GRCh38
rs969998845 Chr.4: 673367-673367 on Build GRCh38
rs755880121 Chr.4: 673310-673320 on Build GRCh38
rs778726707 Chr.4: 673418-673420 on Build GRCh38
rs769709435 Chr.4: 673332-673332 on Build GRCh38
rs902011351 Chr.4: 673323-673323 on Build GRCh38
rs775275554 Chr.4: 673263-673263 on Build GRCh38
rs920052741 Chr.4: 673389-673389 on Build GRCh38
rs943009816 Chr.4: 673464-673464 on Build GRCh38
rs373609558 Chr.4: 673411-673411 on Build GRCh38
rs762196712 Chr.4: 673436-673436 on Build GRCh38
rs201986374 Chr.4: 673294-673294 on Build GRCh38
rs761788482 Chr.4: 673272-673272 on Build GRCh38
rs150476333 Chr.4: 673308-673308 on Build GRCh38
rs113061105 Chr.4: 673302-673302 on Build GRCh38
rs80010815 Chr.4: 673287-673287 on Build GRCh38
rs534828673 Chr.4: 673352-673352 on Build GRCh38
rs772776063 Chr.4: 673280-673280 on Build GRCh38
rs980599228 Chr.4: 673373-673373 on Build GRCh38
rs149611704 Chr.4: 673338-673338 on Build GRCh38
rs774192635 Chr.4: 673346-673346 on Build GRCh38
rs375236762 Chr.4: 673349-673349 on Build GRCh38
rs201475846 Chr.4: 673449-673449 on Build GRCh38
rs879234770 Chr.4: 673402-673402 on Build GRCh38
rs1008217134 Chr.4: 673444-673444 on Build GRCh38
rs138351443 Chr.4: 673328-673328 on Build GRCh38
rs776591346 Chr.4: 673271-673271 on Build GRCh38
rs747922163 Chr.4: 673386-673386 on Build GRCh38
rs777663450 Chr.4: 673337-673364 on Build GRCh38
rs778859823 Chr.4: 673298-673298 on Build GRCh38
rs375900377 Chr.4: 673400-673400 on Build GRCh38
rs762238802 Chr.4: 673350-673350 on Build GRCh38
rs757168270 Chr.4: 673388-673394 on Build GRCh38
rs371350754 Chr.4: 673270-673270 on Build GRCh38
rs774911734 Chr.4: 673248-673248 on Build GRCh38
rs755191067 Chr.4: 673383-673383 on Build GRCh38
rs772213760 Chr.4: 673254-673254 on Build GRCh38
rs773990627 Chr.4: 673410-673410 on Build GRCh38
rs754356649 Chr.4: 673289-673289 on Build GRCh38
rs765723995 Chr.4: 673354-673354 on Build GRCh38
rs748101684 Chr.4: 673331-673331 on Build GRCh38
rs753605745 Chr.4: 673343-673345 on Build GRCh38
rs147287885 Chr.4: 673345-673345 on Build GRCh38
rs552730696 Chr.4: 673416-673416 on Build GRCh38
rs780209628 Chr.4: 673319-673319 on Build GRCh38
rs777730658 Chr.4: 673396-673396 on Build GRCh38
rs751837855 Chr.4: 673377-673377 on Build GRCh38
rs112862871 Chr.4: 673402-673402 on Build GRCh38
rs750500007 Chr.4: 673362-673362 on Build GRCh38
rs769643449 Chr.4: 673394-673394 on Build GRCh38
rs181881252 Chr.4: 673326-673326 on Build GRCh38
rs752985057 Chr.4: 673287-673287 on Build GRCh38
rs746757962 Chr.4: 673268-673268 on Build GRCh38
rs770620537 Chr.4: 673401-673401 on Build GRCh38

View Full Product Details