Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66492923-66493150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEAN1
Entrez Gene ID
146227
Gene Name
brain expressed, associated with NEDD4, 1
Gene Aliases
BEAN, SCA31
UniGene
Hs.740218
Gene Chromosome Location
Chr.16: 66427297-66495288 on Build GRCh38
Transcripts
NM_001197224.3 NM_001197225.3

SNP Detail

SNP ID Chromosome Location
rs915400941 Chr.16: 66493121-66493121 on Build GRCh38
rs1029757809 Chr.16: 66492946-66492946 on Build GRCh38
rs560803672 Chr.16: 66493034-66493034 on Build GRCh38
rs1005892328 Chr.16: 66492969-66492969 on Build GRCh38
rs751786635 Chr.16: 66492972-66492972 on Build GRCh38
rs971699304 Chr.16: 66493020-66493020 on Build GRCh38
rs754915917 Chr.16: 66493030-66493030 on Build GRCh38
rs763587014 Chr.16: 66493022-66493022 on Build GRCh38
rs951401590 Chr.16: 66492992-66492992 on Build GRCh38
rs1015973076 Chr.16: 66492999-66492999 on Build GRCh38
rs150475902 Chr.16: 66493064-66493064 on Build GRCh38
rs754537210 Chr.16: 66493130-66493130 on Build GRCh38
rs911363927 Chr.16: 66493066-66493066 on Build GRCh38
rs189628750 Chr.16: 66493047-66493047 on Build GRCh38
rs766221952 Chr.16: 66493009-66493009 on Build GRCh38
rs971079701 Chr.16: 66493037-66493037 on Build GRCh38
rs540866817 Chr.16: 66493021-66493021 on Build GRCh38
rs755809041 Chr.16: 66492980-66492980 on Build GRCh38
rs181104683 Chr.16: 66493049-66493049 on Build GRCh38
rs185256699 Chr.16: 66493077-66493077 on Build GRCh38
rs988282843 Chr.16: 66493112-66493112 on Build GRCh38
rs990194172 Chr.16: 66493078-66493078 on Build GRCh38
rs961811673 Chr.16: 66493012-66493012 on Build GRCh38
rs764296672 Chr.16: 66492961-66492961 on Build GRCh38
rs962978506 Chr.16: 66493001-66493001 on Build GRCh38
rs780158769 Chr.16: 66492993-66492993 on Build GRCh38

View Full Product Details