Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 102878030-102878263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL11A1
Entrez Gene ID
1301
Gene Name
collagen type XI alpha 1 chain
Gene Aliases
CO11A1, COLL6, STL2
UniGene
Hs.523446
Gene Chromosome Location
Chr.1: 102876467-103108580 on Build GRCh38
Transcripts
NM_001190709.1 NM_001854.3 NM_080629.2 NM_080630.3 NR_134980.1 XM_017000334.1 XM_017000335.1 XM_017000337.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7241913 c.5336C>T p.P1779L Chr.1: 102878104-102878104 on Build GRCh38
6533795 c.5319A>C p.K1773N Chr.1: 102878157-102878157 on Build GRCh38
4019950 c.5398T>G p.F1800V Chr.1: 102878078-102878078 on Build GRCh38
1626257 c.5417A>T p.K1806M Chr.1: 102878059-102878059 on Build GRCh38
6197018 c.5364T>A p.D1788E Chr.1: 102878112-102878112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200514622 Chr.1: 102878210-102878210 on Build GRCh38
rs748518624 Chr.1: 102878180-102878180 on Build GRCh38
rs199848923 Chr.1: 102878126-102878126 on Build GRCh38
rs775083694 Chr.1: 102878093-102878093 on Build GRCh38
rs571497194 Chr.1: 102878078-102878078 on Build GRCh38
rs770392400 Chr.1: 102878055-102878055 on Build GRCh38
rs756303349 Chr.1: 102878148-102878148 on Build GRCh38
rs755371727 Chr.1: 102878168-102878168 on Build GRCh38
rs991011471 Chr.1: 102878108-102878108 on Build GRCh38
rs367853741 Chr.1: 102878165-102878165 on Build GRCh38
rs144510951 Chr.1: 102878081-102878081 on Build GRCh38
rs925433052 Chr.1: 102878054-102878054 on Build GRCh38
rs1036630733 Chr.1: 102878136-102878136 on Build GRCh38
rs200949243 Chr.1: 102878174-102878174 on Build GRCh38
rs777439432 Chr.1: 102878193-102878193 on Build GRCh38
rs746621015 Chr.1: 102878195-102878195 on Build GRCh38
rs750652823 Chr.1: 102878123-102878123 on Build GRCh38
rs930447176 Chr.1: 102878219-102878219 on Build GRCh38
rs372652686 Chr.1: 102878191-102878191 on Build GRCh38
rs949030111 Chr.1: 102878196-102878196 on Build GRCh38
rs762806617 Chr.1: 102878111-102878111 on Build GRCh38
rs532113782 Chr.1: 102878115-102878115 on Build GRCh38
rs765097661 Chr.1: 102878085-102878085 on Build GRCh38
rs940451517 Chr.1: 102878084-102878084 on Build GRCh38
rs766529064 Chr.1: 102878156-102878156 on Build GRCh38

View Full Product Details