Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 90308771-90308998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCSER1
Entrez Gene ID
401145
Gene Name
coiled-coil serine rich protein 1
Gene Aliases
FAM190A
UniGene
Hs.654735
Gene Chromosome Location
Chr.4: 90127394-91602219 on Build GRCh38
Transcripts
NM_001145065.1 NM_207491.2 XM_011531936.1 XM_011531937.2 XM_011531938.2 XM_011531939.2 XM_011531941.2 XM_011531942.2 XM_011531943.2 XM_011531945.1 XM_011531946.2 XM_011531947.2 XM_011531948.1 XM_011531949.2 XM_011531950.2 XM_011531951.2 XM_011531955.2 XM_011531956.2 XM_011531957.2 XM_011531958.2 XM_017008194.1 XM_017008195.1 XM_017008196.1 XM_017008197.1 XM_017008198.1 XM_017008199.1 XM_017008200.1 XM_017008201.1 XM_017008202.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7252645 c.713C>T p.A238V Chr.4: 90308997-90308997 on Build GRCh38
5183902 c.681G>A p.S227S Chr.4: 90308965-90308965 on Build GRCh38
6198687 c.512G>A p.R171H Chr.4: 90308796-90308796 on Build GRCh38
735310 c.583G>T p.V195F Chr.4: 90308867-90308867 on Build GRCh38
1134213 c.643C>T p.Q215* Chr.4: 90308927-90308927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549014692 Chr.4: 90308793-90308793 on Build GRCh38
rs747459246 Chr.4: 90308944-90308944 on Build GRCh38
rs1017686730 Chr.4: 90308869-90308869 on Build GRCh38
rs1034101392 Chr.4: 90308963-90308963 on Build GRCh38
rs751692185 Chr.4: 90308973-90308973 on Build GRCh38
rs766678804 Chr.4: 90308815-90308815 on Build GRCh38
rs964442979 Chr.4: 90308883-90308883 on Build GRCh38
rs781070134 Chr.4: 90308898-90308898 on Build GRCh38
rs748810862 Chr.4: 90308796-90308796 on Build GRCh38
rs773629286 Chr.4: 90308808-90308808 on Build GRCh38
rs775534176 Chr.4: 90308961-90308961 on Build GRCh38
rs759686808 Chr.4: 90308822-90308822 on Build GRCh38
rs954453775 Chr.4: 90308812-90308812 on Build GRCh38
rs929452063 Chr.4: 90308943-90308943 on Build GRCh38
rs748805446 Chr.4: 90308926-90308926 on Build GRCh38
rs764266861 Chr.4: 90308842-90308842 on Build GRCh38
rs766030400 Chr.4: 90308849-90308850 on Build GRCh38
rs776992168 Chr.4: 90308817-90308817 on Build GRCh38
rs751405145 Chr.4: 90308865-90308865 on Build GRCh38
rs771434691 Chr.4: 90308950-90308950 on Build GRCh38
rs761199208 Chr.4: 90308814-90308814 on Build GRCh38
rs868728582 Chr.4: 90308884-90308884 on Build GRCh38
rs199696987 Chr.4: 90308795-90308795 on Build GRCh38
rs757275965 Chr.4: 90308890-90308890 on Build GRCh38
rs759952569 Chr.4: 90308955-90308955 on Build GRCh38
rs752808705 Chr.4: 90308835-90308835 on Build GRCh38
rs764021572 Chr.4: 90308965-90308965 on Build GRCh38
rs903950516 Chr.4: 90308831-90308831 on Build GRCh38
rs202076608 Chr.4: 90308915-90308915 on Build GRCh38
rs538262643 Chr.4: 90308929-90308929 on Build GRCh38
rs770035676 Chr.4: 90308960-90308960 on Build GRCh38
rs776976309 Chr.4: 90308954-90308954 on Build GRCh38
rs750313546 Chr.4: 90308904-90308904 on Build GRCh38
rs974051025 Chr.4: 90308921-90308921 on Build GRCh38
rs763258132 Chr.4: 90308964-90308964 on Build GRCh38

View Full Product Details