Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111495092-111495365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG4
Entrez Gene ID
54766
Gene Name
BTG anti-proliferation factor 4
Gene Aliases
APRO3, PC3B
UniGene
Hs.128180
Gene Chromosome Location
Chr.11: 111383828-111512388 on Build GRCh38
Transcripts
NM_017589.3 XM_011542876.2 XM_011542878.2 XM_011542879.2 XM_011542880.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1246310 c.576C>T p.D192D Chr.11: 111495249-111495249 on Build GRCh38
5371635 c.564G>A p.K188K Chr.11: 111495261-111495261 on Build GRCh38
5341402 c.594G>C p.L198L Chr.11: 111495231-111495231 on Build GRCh38
5506380 c.511-5delT p.? Chr.11: 111495319-111495319 on Build GRCh38
1351205 c.662G>T p.R221I Chr.11: 111495163-111495163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201313198 Chr.11: 111495244-111495244 on Build GRCh38
rs1044746283 Chr.11: 111495257-111495257 on Build GRCh38
rs764389963 Chr.11: 111495127-111495127 on Build GRCh38
rs369066698 Chr.11: 111495223-111495223 on Build GRCh38
rs777187788 Chr.11: 111495211-111495211 on Build GRCh38
rs144985096 Chr.11: 111495140-111495140 on Build GRCh38
rs61751528 Chr.11: 111495318-111495318 on Build GRCh38
rs747532157 Chr.11: 111495282-111495282 on Build GRCh38
rs749138082 Chr.11: 111495266-111495266 on Build GRCh38
rs776579312 Chr.11: 111495129-111495129 on Build GRCh38
rs933415825 Chr.11: 111495245-111495245 on Build GRCh38
rs769105147 Chr.11: 111495300-111495300 on Build GRCh38
rs367799263 Chr.11: 111495119-111495119 on Build GRCh38
rs771189683 Chr.11: 111495145-111495145 on Build GRCh38
rs761044453 Chr.11: 111495125-111495125 on Build GRCh38
rs567920376 Chr.11: 111495139-111495139 on Build GRCh38
rs574289357 Chr.11: 111495158-111495158 on Build GRCh38
rs200079528 Chr.11: 111495268-111495268 on Build GRCh38
rs746112390 Chr.11: 111495272-111495272 on Build GRCh38
rs750056268 Chr.11: 111495128-111495128 on Build GRCh38
rs533540949 Chr.11: 111495236-111495236 on Build GRCh38
rs888215078 Chr.11: 111495217-111495217 on Build GRCh38
rs113732510 Chr.11: 111495185-111495185 on Build GRCh38
rs768629922 Chr.11: 111495318-111495319 on Build GRCh38
rs763061244 Chr.11: 111495118-111495118 on Build GRCh38
rs115316165 Chr.11: 111495134-111495134 on Build GRCh38
rs763997236 Chr.11: 111495135-111495135 on Build GRCh38
rs766954448 Chr.11: 111495248-111495248 on Build GRCh38
rs765708783 Chr.11: 111495321-111495321 on Build GRCh38
rs201605244 Chr.11: 111495259-111495259 on Build GRCh38
rs766203310 Chr.11: 111495234-111495234 on Build GRCh38
rs989099560 Chr.11: 111495174-111495174 on Build GRCh38
rs774094433 Chr.11: 111495325-111495325 on Build GRCh38
rs150474228 Chr.11: 111495279-111495279 on Build GRCh38
rs762204595 Chr.11: 111495316-111495316 on Build GRCh38
rs1003759943 Chr.11: 111495262-111495262 on Build GRCh38
rs754863791 Chr.11: 111495137-111495137 on Build GRCh38
rs957095657 Chr.11: 111495115-111495115 on Build GRCh38
rs761118451 Chr.11: 111495197-111495197 on Build GRCh38
rs918385029 Chr.11: 111495186-111495186 on Build GRCh38
rs761523046 Chr.11: 111495322-111495322 on Build GRCh38
rs140812937 Chr.11: 111495269-111495269 on Build GRCh38
rs759138273 Chr.11: 111495330-111495330 on Build GRCh38
rs549155500 Chr.11: 111495314-111495314 on Build GRCh38
rs376895795 Chr.11: 111495144-111495144 on Build GRCh38
rs772058893 Chr.11: 111495173-111495173 on Build GRCh38
rs764491994 Chr.11: 111495210-111495210 on Build GRCh38
rs772151188 Chr.11: 111495273-111495273 on Build GRCh38
rs781301525 Chr.11: 111495319-111495319 on Build GRCh38
rs767021449 Chr.11: 111495333-111495333 on Build GRCh38
rs752636903 Chr.11: 111495249-111495249 on Build GRCh38
rs137858975 Chr.11: 111495179-111495179 on Build GRCh38
rs765968342 Chr.11: 111495133-111495133 on Build GRCh38
rs756046366 Chr.11: 111495255-111495255 on Build GRCh38
rs1031678834 Chr.11: 111495265-111495265 on Build GRCh38
rs544902747 Chr.11: 111495172-111495172 on Build GRCh38

View Full Product Details