Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74474499-74474740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300A
Entrez Gene ID
11314
Gene Name
CD300a molecule
Gene Aliases
CLM-8, CMRF-35-H9, CMRF-35H, CMRF35-H, CMRF35-H9, CMRF35H, CMRF35H9, IGSF12, IRC1, IRC1/IRC2, IRC2, IRp60
UniGene
Hs.9688
Gene Chromosome Location
Chr.17: 74466320-74484798 on Build GRCh38
Transcripts
NM_001256841.1 NM_007261.3 XM_005256990.2 XM_005256991.4 XM_005256992.4 XM_006721656.3 XM_011524254.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
984059 c.422C>T p.T141I Chr.17: 74474574-74474574 on Build GRCh38
6480892 c.513T>A p.T171T Chr.17: 74474665-74474665 on Build GRCh38
984060 c.430A>C p.I144L Chr.17: 74474582-74474582 on Build GRCh38
6532552 c.521T>C p.V174A Chr.17: 74474673-74474673 on Build GRCh38
437320 c.388T>A p.S130T Chr.17: 74474540-74474540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756681723 Chr.17: 74474576-74474576 on Build GRCh38
rs775254373 Chr.17: 74474647-74474647 on Build GRCh38
rs774342665 Chr.17: 74474672-74474672 on Build GRCh38
rs776573991 Chr.17: 74474656-74474656 on Build GRCh38
rs1018422074 Chr.17: 74474521-74474521 on Build GRCh38
rs7208061 Chr.17: 74474700-74474700 on Build GRCh38
rs755193796 Chr.17: 74474559-74474559 on Build GRCh38
rs746874556 Chr.17: 74474654-74474654 on Build GRCh38
rs754866896 Chr.17: 74474715-74474715 on Build GRCh38
rs373628688 Chr.17: 74474543-74474543 on Build GRCh38
rs779966637 Chr.17: 74474713-74474714 on Build GRCh38
rs751730343 Chr.17: 74474518-74474518 on Build GRCh38
rs200630749 Chr.17: 74474678-74474678 on Build GRCh38
rs761687016 Chr.17: 74474664-74474664 on Build GRCh38
rs139953037 Chr.17: 74474565-74474565 on Build GRCh38
rs753800485 Chr.17: 74474579-74474579 on Build GRCh38
rs764579323 Chr.17: 74474709-74474709 on Build GRCh38
rs377134558 Chr.17: 74474550-74474550 on Build GRCh38
rs750498629 Chr.17: 74474538-74474538 on Build GRCh38
rs749768413 Chr.17: 74474586-74474586 on Build GRCh38
rs770430150 Chr.17: 74474590-74474590 on Build GRCh38
rs778353917 Chr.17: 74474577-74474577 on Build GRCh38
rs976988186 Chr.17: 74474553-74474553 on Build GRCh38
rs753167623 Chr.17: 74474569-74474569 on Build GRCh38
rs201023984 Chr.17: 74474532-74474532 on Build GRCh38
rs766450811 Chr.17: 74474671-74474671 on Build GRCh38
rs1027136152 Chr.17: 74474692-74474692 on Build GRCh38
rs754333758 Chr.17: 74474712-74474712 on Build GRCh38
rs1008144753 Chr.17: 74474566-74474566 on Build GRCh38
rs745506428 Chr.17: 74474615-74474615 on Build GRCh38
rs757748407 Chr.17: 74474718-74474718 on Build GRCh38
rs778321916 Chr.17: 74474593-74474593 on Build GRCh38
rs771763718 Chr.17: 74474640-74474640 on Build GRCh38
rs369450007 Chr.17: 74474539-74474539 on Build GRCh38
rs752969166 Chr.17: 74474691-74474691 on Build GRCh38

View Full Product Details