Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85212053-85212326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F5
Entrez Gene ID
1875
Gene Name
E2F transcription factor 5
Gene Aliases
E2F-5
UniGene
Hs.445758
Gene Chromosome Location
Chr.8: 85177384-85214518 on Build GRCh38
Transcripts
NM_001083588.1 NM_001083589.1 NM_001951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4151908 c.884-3C>T p.? Chr.8: 85212154-85212154 on Build GRCh38
1330893 c.908T>C p.I303T Chr.8: 85212181-85212181 on Build GRCh38
2877362 c.899G>A p.G300E Chr.8: 85212172-85212172 on Build GRCh38
7150804 c.906C>G p.I302M Chr.8: 85212179-85212179 on Build GRCh38
6114231 c.906C>T p.I302I Chr.8: 85212179-85212179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018624328 Chr.8: 85212266-85212266 on Build GRCh38
rs996199823 Chr.8: 85212139-85212139 on Build GRCh38
rs758658729 Chr.8: 85212082-85212083 on Build GRCh38
rs771884834 Chr.8: 85212246-85212246 on Build GRCh38
rs142778143 Chr.8: 85212077-85212077 on Build GRCh38
rs749464145 Chr.8: 85212235-85212235 on Build GRCh38
rs189297065 Chr.8: 85212207-85212207 on Build GRCh38
rs745737622 Chr.8: 85212244-85212244 on Build GRCh38
rs755426627 Chr.8: 85212203-85212203 on Build GRCh38
rs889979856 Chr.8: 85212181-85212181 on Build GRCh38
rs756223877 Chr.8: 85212210-85212210 on Build GRCh38
rs761634364 Chr.8: 85212260-85212260 on Build GRCh38
rs752791569 Chr.8: 85212208-85212208 on Build GRCh38
rs757507239 Chr.8: 85212240-85212240 on Build GRCh38
rs769707076 Chr.8: 85212191-85212191 on Build GRCh38
rs1049902397 Chr.8: 85212141-85212141 on Build GRCh38
rs1037043893 Chr.8: 85212085-85212085 on Build GRCh38
rs779296136 Chr.8: 85212135-85212136 on Build GRCh38
rs943595547 Chr.8: 85212104-85212104 on Build GRCh38
rs780010292 Chr.8: 85212252-85212252 on Build GRCh38
rs574493647 Chr.8: 85212297-85212297 on Build GRCh38
rs368647550 Chr.8: 85212177-85212177 on Build GRCh38
rs552106113 Chr.8: 85212134-85212134 on Build GRCh38
rs753974669 Chr.8: 85212118-85212119 on Build GRCh38
rs899335859 Chr.8: 85212169-85212169 on Build GRCh38
rs778571444 Chr.8: 85212243-85212243 on Build GRCh38
rs776214566 Chr.8: 85212131-85212131 on Build GRCh38
rs868730092 Chr.8: 85212204-85212204 on Build GRCh38
rs755198174 Chr.8: 85212124-85212124 on Build GRCh38
rs766581402 Chr.8: 85212135-85212135 on Build GRCh38
rs1047454731 Chr.8: 85212249-85212249 on Build GRCh38
rs372107449 Chr.8: 85212218-85212218 on Build GRCh38
rs752024109 Chr.8: 85212163-85212163 on Build GRCh38
rs778022827 Chr.8: 85212212-85212212 on Build GRCh38

View Full Product Details