Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185399431-185399664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD37
Entrez Gene ID
353322
Gene Name
ankyrin repeat domain 37
Gene Aliases
Lrp2bp
UniGene
Hs.508154
Gene Chromosome Location
Chr.4: 185396686-185400245 on Build GRCh38
Transcripts
NM_181726.3 XM_017008176.1 NM_018359.3 NR_028085.1

Gene Symbol
UFSP2
Entrez Gene ID
55325
Gene Name
UFM1 specific peptidase 2
Gene Aliases
BHD, C4orf20
UniGene
Hs.713548
Gene Chromosome Location
Chr.4: 185399540-185425985 on Build GRCh38
Transcripts
NM_181726.3 XM_017008176.1 NM_018359.3 NR_028085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4820534 c.304G>A p.D102N Chr.4: 185399601-185399601 on Build GRCh38
4429941 c.276A>G p.L92L Chr.4: 185399573-185399573 on Build GRCh38
3392804 c.297A>G p.T99T Chr.4: 185399594-185399594 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754163591 Chr.4: 185399611-185399611 on Build GRCh38
rs543985284 Chr.4: 185399504-185399504 on Build GRCh38
rs202213765 Chr.4: 185399513-185399513 on Build GRCh38
rs757623071 Chr.4: 185399614-185399614 on Build GRCh38
rs747977169 Chr.4: 185399561-185399561 on Build GRCh38
rs774437070 Chr.4: 185399574-185399574 on Build GRCh38
rs768637598 Chr.4: 185399522-185399522 on Build GRCh38
rs776541906 Chr.4: 185399559-185399559 on Build GRCh38
rs397701725 Chr.4: 185399521-185399521 on Build GRCh38
rs1049990788 Chr.4: 185399606-185399606 on Build GRCh38
rs188327719 Chr.4: 185399562-185399562 on Build GRCh38
rs759865037 Chr.4: 185399582-185399582 on Build GRCh38
rs992186883 Chr.4: 185399515-185399515 on Build GRCh38
rs926898313 Chr.4: 185399512-185399512 on Build GRCh38
rs1027249949 Chr.4: 185399578-185399578 on Build GRCh38
rs932501762 Chr.4: 185399596-185399596 on Build GRCh38
rs11287696 Chr.4: 185399513-185399513 on Build GRCh38
rs961213586 Chr.4: 185399514-185399514 on Build GRCh38
rs760164904 Chr.4: 185399622-185399622 on Build GRCh38
rs368868442 Chr.4: 185399585-185399585 on Build GRCh38
rs768752275 Chr.4: 185399542-185399542 on Build GRCh38
rs761152223 Chr.4: 185399598-185399598 on Build GRCh38
rs764886919 Chr.4: 185399626-185399626 on Build GRCh38
rs375912914 Chr.4: 185399607-185399607 on Build GRCh38
rs779672312 Chr.4: 185399527-185399527 on Build GRCh38
rs981346100 Chr.4: 185399473-185399473 on Build GRCh38
rs201802691 Chr.4: 185399586-185399586 on Build GRCh38
rs750030790 Chr.4: 185399631-185399631 on Build GRCh38
rs374431786 Chr.4: 185399534-185399534 on Build GRCh38
rs757860953 Chr.4: 185399522-185399522 on Build GRCh38
rs760381441 Chr.4: 185399500-185399500 on Build GRCh38

View Full Product Details