Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 29018347-29018572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
UniGene
Hs.175437
Gene Chromosome Location
Chr.1: 28887091-29120046 on Build GRCh38
Transcripts
NM_001166005.1 NM_001166006.1 NM_001166007.1 NM_004437.3 NM_203342.2 NM_203343.2 XM_005245753.1 XM_005245757.1 XM_005245760.1 XM_005245761.1 XM_005245763.1 XM_005245764.1 XM_005245765.1 XM_005245768.1 XM_005245769.1 XM_005245770.1 XM_005245772.4 XM_005245773.4 XM_005245774.1 XM_006710434.1 XM_006710439.1 XM_011540956.1 XM_011540957.1 XM_011540958.1 XM_011540959.1 XM_011540960.1 XM_011540961.1 XM_011540962.1 XM_011540963.2 XM_011540964.1 XM_011540965.2 XM_017000581.1 XM_017000582.1 XM_017000583.1 XM_017000584.1 XM_017000585.1 XM_017000586.1 XM_017000587.1 XM_017000588.1 XM_017000589.1 XM_017000590.1 XM_017000591.1 XM_017000592.1 XM_017000593.1 XM_017000594.1 XM_017000595.1 XM_017000596.1 XM_017000597.1 XM_017000598.1 XM_017000599.1 XM_017000600.1 XM_017000601.1 XM_017000602.1 XM_017000603.1 XM_017000604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363263 c.1039G>T p.V347L Chr.1: 29018357-29018357 on Build GRCh38
6363264 c.412G>T p.V138L Chr.1: 29018357-29018357 on Build GRCh38
5685549 c.485A>G p.H162R Chr.1: 29018430-29018430 on Build GRCh38
7428414 c.444G>A p.P148P Chr.1: 29018389-29018389 on Build GRCh38
5993152 c.1038C>T p.G346G Chr.1: 29018356-29018356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs33976881 Chr.1: 29018498-29018499 on Build GRCh38
rs56042538 Chr.1: 29018496-29018496 on Build GRCh38
rs201817969 Chr.1: 29018498-29018498 on Build GRCh38
rs1054714853 Chr.1: 29018435-29018435 on Build GRCh38
rs869025285 Chr.1: 29018389-29018395 on Build GRCh38
rs773724136 Chr.1: 29018484-29018484 on Build GRCh38
rs755228773 Chr.1: 29018392-29018392 on Build GRCh38
rs999513386 Chr.1: 29018469-29018469 on Build GRCh38
rs55946958 Chr.1: 29018497-29018497 on Build GRCh38
rs751756531 Chr.1: 29018391-29018391 on Build GRCh38
rs72508794 Chr.1: 29018496-29018497 on Build GRCh38
rs867840557 Chr.1: 29018525-29018525 on Build GRCh38
rs758164060 Chr.1: 29018443-29018443 on Build GRCh38
rs377224137 Chr.1: 29018467-29018467 on Build GRCh38
rs372904193 Chr.1: 29018454-29018454 on Build GRCh38
rs1012748776 Chr.1: 29018440-29018440 on Build GRCh38
rs766661378 Chr.1: 29018467-29018467 on Build GRCh38
rs771374620 Chr.1: 29018491-29018491 on Build GRCh38
rs202197419 Chr.1: 29018388-29018388 on Build GRCh38
rs911979168 Chr.1: 29018479-29018479 on Build GRCh38
rs763248610 Chr.1: 29018487-29018487 on Build GRCh38
rs143958458 Chr.1: 29018413-29018413 on Build GRCh38
rs571455233 Chr.1: 29018514-29018514 on Build GRCh38
rs369739885 Chr.1: 29018483-29018483 on Build GRCh38
rs2985334 Chr.1: 29018515-29018515 on Build GRCh38
rs781555462 Chr.1: 29018508-29018508 on Build GRCh38
rs143506647 Chr.1: 29018482-29018482 on Build GRCh38
rs200261627 Chr.1: 29018422-29018422 on Build GRCh38
rs372873942 Chr.1: 29018434-29018434 on Build GRCh38
rs778928272 Chr.1: 29018430-29018430 on Build GRCh38
rs201509659 Chr.1: 29018389-29018389 on Build GRCh38
rs746718791 Chr.1: 29018464-29018464 on Build GRCh38
rs200348830 Chr.1: 29018499-29018499 on Build GRCh38
rs5773229 Chr.1: 29018497-29018498 on Build GRCh38
rs752818050 Chr.1: 29018419-29018419 on Build GRCh38
rs770303414 Chr.1: 29018542-29018542 on Build GRCh38

View Full Product Details