Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143867637-143867882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPPK1
Entrez Gene ID
83481
Gene Name
epiplakin 1
Gene Aliases
EPIPL, EPIPL1
UniGene
Hs.200412
Gene Chromosome Location
Chr.8: 143857319-143879043 on Build GRCh38
Transcripts
NM_031308.3 XM_017013889.1 XM_017013890.1 XM_017013891.1 XM_017013892.1

SNP Detail

SNP ID Chromosome Location
rs782367327 Chr.8: 143867849-143867849 on Build GRCh38
rs781870609 Chr.8: 143867715-143867715 on Build GRCh38
rs370265526 Chr.8: 143867823-143867823 on Build GRCh38
rs373276585 Chr.8: 143867775-143867775 on Build GRCh38
rs371108096 Chr.8: 143867821-143867821 on Build GRCh38
rs376434550 Chr.8: 143867816-143867816 on Build GRCh38
rs782728453 Chr.8: 143867803-143867803 on Build GRCh38
rs781928589 Chr.8: 143867770-143867770 on Build GRCh38
rs782233446 Chr.8: 143867829-143867829 on Build GRCh38
rs782698531 Chr.8: 143867820-143867820 on Build GRCh38
rs782776705 Chr.8: 143867809-143867809 on Build GRCh38
rs561337449 Chr.8: 143867846-143867846 on Build GRCh38
rs372409375 Chr.8: 143867857-143867857 on Build GRCh38
rs897305222 Chr.8: 143867757-143867757 on Build GRCh38
rs376518639 Chr.8: 143867787-143867787 on Build GRCh38
rs534326544 Chr.8: 143867739-143867739 on Build GRCh38
rs552597500 Chr.8: 143867751-143867751 on Build GRCh38
rs782160316 Chr.8: 143867710-143867710 on Build GRCh38
rs377154590 Chr.8: 143867860-143867860 on Build GRCh38
rs554713560 Chr.8: 143867811-143867811 on Build GRCh38
rs782010042 Chr.8: 143867707-143867707 on Build GRCh38
rs150969952 Chr.8: 143867711-143867711 on Build GRCh38
rs782604818 Chr.8: 143867756-143867756 on Build GRCh38
rs200136005 Chr.8: 143867822-143867822 on Build GRCh38
rs1017044924 Chr.8: 143867755-143867755 on Build GRCh38
rs369677374 Chr.8: 143867658-143867658 on Build GRCh38
rs372006372 Chr.8: 143867659-143867659 on Build GRCh38
rs782448102 Chr.8: 143867724-143867724 on Build GRCh38
rs782477671 Chr.8: 143867716-143867716 on Build GRCh38
rs782736381 Chr.8: 143867696-143867696 on Build GRCh38
rs782342687 Chr.8: 143867677-143867677 on Build GRCh38
rs544053812 Chr.8: 143867824-143867824 on Build GRCh38
rs781969807 Chr.8: 143867791-143867791 on Build GRCh38
rs782316359 Chr.8: 143867763-143867763 on Build GRCh38
rs79961029 Chr.8: 143867735-143867735 on Build GRCh38
rs371009588 Chr.8: 143867790-143867790 on Build GRCh38
rs73715514 Chr.8: 143867772-143867772 on Build GRCh38
rs782230047 Chr.8: 143867758-143867758 on Build GRCh38
rs541551993 Chr.8: 143867806-143867806 on Build GRCh38
rs781935385 Chr.8: 143867844-143867844 on Build GRCh38
rs781976496 Chr.8: 143867687-143867687 on Build GRCh38
rs782563487 Chr.8: 143867815-143867815 on Build GRCh38
rs782735649 Chr.8: 143867719-143867719 on Build GRCh38
rs782139638 Chr.8: 143867782-143867782 on Build GRCh38
rs200992058 Chr.8: 143867797-143867797 on Build GRCh38
rs782484203 Chr.8: 143867812-143867812 on Build GRCh38
rs782054117 Chr.8: 143867694-143867694 on Build GRCh38
rs782521746 Chr.8: 143867817-143867817 on Build GRCh38
rs374536102 Chr.8: 143867842-143867842 on Build GRCh38
rs373216268 Chr.8: 143867723-143867723 on Build GRCh38

View Full Product Details