Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101908147-101908394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRAM1
Entrez Gene ID
55332
Gene Name
DNA damage regulated autophagy modulator 1
Gene Aliases
DRAM
UniGene
Hs.525634
Gene Chromosome Location
Chr.12: 101877327-101923623 on Build GRCh38
Transcripts
NM_018370.2 XM_005269004.2 XM_005269005.2 XM_017019578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5004328 c.506C>T p.A169V Chr.12: 101908349-101908349 on Build GRCh38
239645 c.193G>C p.V65L Chr.12: 101908354-101908354 on Build GRCh38
302097 c.40G>A p.V14M Chr.12: 101908201-101908201 on Build GRCh38
5373973 c.393C>T p.V131V Chr.12: 101908236-101908236 on Build GRCh38
3383974 c.160C>T p.R54W Chr.12: 101908321-101908321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745515371 Chr.12: 101908285-101908285 on Build GRCh38
rs201328131 Chr.12: 101908347-101908347 on Build GRCh38
rs61748065 Chr.12: 101908245-101908245 on Build GRCh38
rs757909615 Chr.12: 101908366-101908366 on Build GRCh38
rs777525199 Chr.12: 101908299-101908299 on Build GRCh38
rs764208584 Chr.12: 101908183-101908183 on Build GRCh38
rs749800547 Chr.12: 101908348-101908348 on Build GRCh38
rs756525009 Chr.12: 101908248-101908248 on Build GRCh38
rs761749222 Chr.12: 101908314-101908314 on Build GRCh38
rs749818728 Chr.12: 101908254-101908254 on Build GRCh38
rs755160311 Chr.12: 101908337-101908337 on Build GRCh38
rs763989214 Chr.12: 101908173-101908173 on Build GRCh38
rs919374528 Chr.12: 101908238-101908238 on Build GRCh38
rs973503827 Chr.12: 101908210-101908210 on Build GRCh38
rs372136588 Chr.12: 101908338-101908338 on Build GRCh38
rs766432965 Chr.12: 101908323-101908323 on Build GRCh38
rs751719297 Chr.12: 101908219-101908219 on Build GRCh38
rs376602463 Chr.12: 101908258-101908258 on Build GRCh38
rs748500198 Chr.12: 101908246-101908246 on Build GRCh38
rs367592764 Chr.12: 101908305-101908305 on Build GRCh38
rs770803171 Chr.12: 101908329-101908329 on Build GRCh38
rs774066101 Chr.12: 101908284-101908284 on Build GRCh38
rs571859082 Chr.12: 101908217-101908217 on Build GRCh38
rs776480486 Chr.12: 101908311-101908311 on Build GRCh38
rs771666007 Chr.12: 101908294-101908294 on Build GRCh38
rs374445581 Chr.12: 101908339-101908339 on Build GRCh38
rs557287563 Chr.12: 101908307-101908307 on Build GRCh38
rs749347997 Chr.12: 101908288-101908288 on Build GRCh38
rs771823620 Chr.12: 101908253-101908253 on Build GRCh38
rs758430600 Chr.12: 101908213-101908213 on Build GRCh38
rs528322922 Chr.12: 101908212-101908212 on Build GRCh38
rs770877289 Chr.12: 101908335-101908344 on Build GRCh38
rs755250610 Chr.12: 101908229-101908229 on Build GRCh38
rs1057443566 Chr.12: 101908270-101908270 on Build GRCh38
rs1050818398 Chr.12: 101908308-101908308 on Build GRCh38

View Full Product Details