Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 124806774-124807013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM175B
Entrez Gene ID
23172
Gene Name
family with sequence similarity 175 member B
Gene Aliases
ABRO1, KIAA0157
UniGene
Hs.280695
Gene Chromosome Location
Chr.10: 124801785-124836670 on Build GRCh38
Transcripts
NM_032182.3 XM_011539554.1 XM_017015986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7149965 c.103G>C p.E35Q Chr.10: 124806861-124806861 on Build GRCh38
4012447 c.74A>G p.E25G Chr.10: 124806832-124806832 on Build GRCh38
6658938 c.89G>A p.G30E Chr.10: 124806847-124806847 on Build GRCh38
260775 c.82T>G p.L28V Chr.10: 124806840-124806840 on Build GRCh38
1346583 c.117C>T p.S39S Chr.10: 124806875-124806875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959877345 Chr.10: 124806849-124806849 on Build GRCh38
rs951349260 Chr.10: 124806895-124806895 on Build GRCh38
rs575121903 Chr.10: 124806915-124806915 on Build GRCh38
rs777440330 Chr.10: 124806911-124806911 on Build GRCh38
rs372729001 Chr.10: 124806851-124806851 on Build GRCh38
rs201425106 Chr.10: 124806949-124806949 on Build GRCh38
rs757954967 Chr.10: 124806860-124806860 on Build GRCh38
rs964311394 Chr.10: 124806935-124806935 on Build GRCh38
rs976100639 Chr.10: 124806964-124806964 on Build GRCh38
rs1034172504 Chr.10: 124806824-124806824 on Build GRCh38
rs758503909 Chr.10: 124806868-124806868 on Build GRCh38
rs759953792 Chr.10: 124806963-124806963 on Build GRCh38
rs764723314 Chr.10: 124806859-124806860 on Build GRCh38
rs772538049 Chr.10: 124806938-124806938 on Build GRCh38
rs779856997 Chr.10: 124806893-124806893 on Build GRCh38
rs771597361 Chr.10: 124806919-124806919 on Build GRCh38
rs752453077 Chr.10: 124806938-124806939 on Build GRCh38
rs1017392254 Chr.10: 124806925-124806925 on Build GRCh38
rs201945744 Chr.10: 124806909-124806909 on Build GRCh38
rs745835758 Chr.10: 124806872-124806872 on Build GRCh38
rs369593747 Chr.10: 124806816-124806816 on Build GRCh38
rs768349234 Chr.10: 124806901-124806901 on Build GRCh38
rs759088149 Chr.10: 124806821-124806825 on Build GRCh38
rs754275177 Chr.10: 124806950-124806950 on Build GRCh38
rs755278032 Chr.10: 124806843-124806843 on Build GRCh38
rs781052243 Chr.10: 124806869-124806869 on Build GRCh38
rs903286601 Chr.10: 124806939-124806939 on Build GRCh38
rs376153017 Chr.10: 124806854-124806854 on Build GRCh38
rs769656894 Chr.10: 124806875-124806875 on Build GRCh38
rs761264467 Chr.10: 124806942-124806942 on Build GRCh38
rs369401224 Chr.10: 124806896-124806896 on Build GRCh38

View Full Product Details