Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37706891-37707162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA8
Entrez Gene ID
55143
Gene Name
cell division cycle associated 8
Gene Aliases
BOR, BOREALIN, DasraB, MESRGP
UniGene
Hs.524571
Gene Chromosome Location
Chr.1: 37692401-37709719 on Build GRCh38
Transcripts
NM_001256875.1 NM_018101.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2167435 c.718C>T p.R240* Chr.1: 37706984-37706984 on Build GRCh38
274462 c.719G>A p.R240Q Chr.1: 37706985-37706985 on Build GRCh38
4506260 c.714C>T p.S238S Chr.1: 37706980-37706980 on Build GRCh38
7120590 c.795C>G p.L265L Chr.1: 37707061-37707061 on Build GRCh38
3977638 c.751A>T p.I251F Chr.1: 37707017-37707017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770826926 Chr.1: 37707093-37707093 on Build GRCh38
rs773989226 Chr.1: 37707101-37707101 on Build GRCh38
rs756051498 Chr.1: 37707079-37707079 on Build GRCh38
rs578232792 Chr.1: 37707134-37707134 on Build GRCh38
rs377582484 Chr.1: 37707047-37707047 on Build GRCh38
rs758729209 Chr.1: 37707034-37707034 on Build GRCh38
rs560264444 Chr.1: 37707056-37707056 on Build GRCh38
rs749016612 Chr.1: 37706946-37706946 on Build GRCh38
rs775841916 Chr.1: 37706959-37706959 on Build GRCh38
rs868020307 Chr.1: 37706994-37706994 on Build GRCh38
rs1017759793 Chr.1: 37706921-37706921 on Build GRCh38
rs754554691 Chr.1: 37707065-37707065 on Build GRCh38
rs369659479 Chr.1: 37707077-37707077 on Build GRCh38
rs536093843 Chr.1: 37706944-37706944 on Build GRCh38
rs776833639 Chr.1: 37706978-37706978 on Build GRCh38
rs779686400 Chr.1: 37706930-37706930 on Build GRCh38
rs757125175 Chr.1: 37707091-37707091 on Build GRCh38
rs777629825 Chr.1: 37707090-37707090 on Build GRCh38
rs191458451 Chr.1: 37707131-37707131 on Build GRCh38
rs780730214 Chr.1: 37707069-37707069 on Build GRCh38
rs764277137 Chr.1: 37706963-37706963 on Build GRCh38
rs45487395 Chr.1: 37707064-37707064 on Build GRCh38
rs748123520 Chr.1: 37706935-37706935 on Build GRCh38
rs148932769 Chr.1: 37706931-37706931 on Build GRCh38
rs868425508 Chr.1: 37707038-37707038 on Build GRCh38
rs747307860 Chr.1: 37707105-37707105 on Build GRCh38
rs183128146 Chr.1: 37706981-37706981 on Build GRCh38
rs780561609 Chr.1: 37706934-37706934 on Build GRCh38
rs765795028 Chr.1: 37706984-37706984 on Build GRCh38
rs750471772 Chr.1: 37706920-37706920 on Build GRCh38
rs760805616 Chr.1: 37706961-37706961 on Build GRCh38
rs1007163560 Chr.1: 37707018-37707018 on Build GRCh38
rs1018352856 Chr.1: 37707061-37707061 on Build GRCh38
rs764229589 Chr.1: 37706948-37706948 on Build GRCh38
rs187658080 Chr.1: 37706985-37706985 on Build GRCh38
rs957886019 Chr.1: 37706987-37706987 on Build GRCh38
rs769696001 Chr.1: 37706937-37706937 on Build GRCh38
rs772199633 Chr.1: 37706951-37706951 on Build GRCh38
rs1046024586 Chr.1: 37706922-37706922 on Build GRCh38

View Full Product Details