Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 172426-172699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOC2B
Entrez Gene ID
8447
Gene Name
double C2 domain beta
Gene Aliases
DOC2BL
UniGene
Hs.551705
Gene Chromosome Location
Chr.17: 146748-181630 on Build GRCh38
Transcripts
NM_003585.4 XM_011535363.2 XM_017025227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4769245 c.386C>T p.T129M Chr.17: 172604-172604 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368833995 Chr.17: 172656-172656 on Build GRCh38
rs1054098698 Chr.17: 172663-172663 on Build GRCh38
rs375087009 Chr.17: 172639-172639 on Build GRCh38
rs1041106119 Chr.17: 172596-172596 on Build GRCh38
rs962050836 Chr.17: 172533-172533 on Build GRCh38
rs899837753 Chr.17: 172603-172603 on Build GRCh38
rs752975986 Chr.17: 172670-172670 on Build GRCh38
rs554935539 Chr.17: 172500-172500 on Build GRCh38
rs544386443 Chr.17: 172585-172585 on Build GRCh38
rs568687375 Chr.17: 172620-172620 on Build GRCh38
rs773549518 Chr.17: 172576-172576 on Build GRCh38
rs148525551 Chr.17: 172517-172517 on Build GRCh38
rs765921136 Chr.17: 172504-172504 on Build GRCh38
rs548235440 Chr.17: 172626-172626 on Build GRCh38
rs200022249 Chr.17: 172615-172615 on Build GRCh38
rs562587513 Chr.17: 172591-172591 on Build GRCh38
rs572918442 Chr.17: 172516-172516 on Build GRCh38
rs565042628 Chr.17: 172518-172518 on Build GRCh38
rs1003162872 Chr.17: 172515-172515 on Build GRCh38
rs773979509 Chr.17: 172528-172528 on Build GRCh38
rs576589254 Chr.17: 172567-172567 on Build GRCh38
rs777813622 Chr.17: 172621-172621 on Build GRCh38
rs778965803 Chr.17: 172644-172644 on Build GRCh38
rs370682502 Chr.17: 172482-172482 on Build GRCh38
rs952108751 Chr.17: 172570-172570 on Build GRCh38
rs1009684677 Chr.17: 172451-172451 on Build GRCh38
rs765879910 Chr.17: 172526-172526 on Build GRCh38
rs772430564 Chr.17: 172573-172575 on Build GRCh38
rs1013047144 Chr.17: 172671-172671 on Build GRCh38
rs751400300 Chr.17: 172527-172527 on Build GRCh38
rs767488468 Chr.17: 172604-172604 on Build GRCh38
rs575894243 Chr.17: 172491-172491 on Build GRCh38
rs989001408 Chr.17: 172640-172640 on Build GRCh38
rs762997281 Chr.17: 172480-172480 on Build GRCh38
rs910679659 Chr.17: 172613-172613 on Build GRCh38
rs536574321 Chr.17: 172496-172496 on Build GRCh38
rs375064949 Chr.17: 172490-172490 on Build GRCh38
rs772740733 Chr.17: 172499-172499 on Build GRCh38
rs894017713 Chr.17: 172447-172447 on Build GRCh38
rs972023376 Chr.17: 172554-172554 on Build GRCh38

View Full Product Details