Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99974365-99974600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS17
Entrez Gene ID
170691
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 17
Gene Aliases
-
UniGene
Hs.513200
Gene Chromosome Location
Chr.15: 99971438-100342924 on Build GRCh38
Transcripts
NM_139057.3 XM_005254872.3 XM_011521312.2 XM_017021973.1 XM_017021975.1 XM_017021976.1 XM_017021978.1 XM_017021979.1 XM_017021980.1 XM_017021982.1 XM_017021983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3499675 c.3175C>T p.R1059W Chr.15: 99974515-99974515 on Build GRCh38
4488097 c.3274C>T p.P1092S Chr.15: 99974416-99974416 on Build GRCh38
6053809 c.3130G>T p.A1044S Chr.15: 99974560-99974560 on Build GRCh38
6055325 c.3192G>T p.K1064N Chr.15: 99974498-99974498 on Build GRCh38
7211355 c.3278C>T p.P1093L Chr.15: 99974412-99974412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148559983 Chr.15: 99974568-99974568 on Build GRCh38
rs201387097 Chr.15: 99974515-99974515 on Build GRCh38
rs755550706 Chr.15: 99974447-99974447 on Build GRCh38
rs777329258 Chr.15: 99974459-99974459 on Build GRCh38
rs372853186 Chr.15: 99974469-99974469 on Build GRCh38
rs371570653 Chr.15: 99974526-99974526 on Build GRCh38
rs757020850 Chr.15: 99974571-99974571 on Build GRCh38
rs886050972 Chr.15: 99974546-99974546 on Build GRCh38
rs773331594 Chr.15: 99974479-99974479 on Build GRCh38
rs766559532 Chr.15: 99974482-99974482 on Build GRCh38
rs777030019 Chr.15: 99974534-99974534 on Build GRCh38
rs780853868 Chr.15: 99974466-99974466 on Build GRCh38
rs1022416989 Chr.15: 99974470-99974470 on Build GRCh38
rs373899380 Chr.15: 99974412-99974412 on Build GRCh38
rs112462169 Chr.15: 99974417-99974417 on Build GRCh38
rs376693261 Chr.15: 99974439-99974439 on Build GRCh38
rs140274211 Chr.15: 99974465-99974465 on Build GRCh38
rs765072526 Chr.15: 99974435-99974435 on Build GRCh38
rs200989311 Chr.15: 99974398-99974398 on Build GRCh38
rs112214202 Chr.15: 99974552-99974552 on Build GRCh38
rs779509724 Chr.15: 99974448-99974448 on Build GRCh38
rs746638782 Chr.15: 99974414-99974414 on Build GRCh38
rs759825365 Chr.15: 99974536-99974536 on Build GRCh38
rs765663952 Chr.15: 99974489-99974489 on Build GRCh38
rs373641684 Chr.15: 99974539-99974539 on Build GRCh38
rs369581271 Chr.15: 99974577-99974577 on Build GRCh38
rs754201774 Chr.15: 99974444-99974444 on Build GRCh38
rs765090779 Chr.15: 99974425-99974425 on Build GRCh38
rs760823694 Chr.15: 99974481-99974481 on Build GRCh38
rs202106289 Chr.15: 99974467-99974467 on Build GRCh38
rs746697133 Chr.15: 99974462-99974462 on Build GRCh38
rs182634994 Chr.15: 99974574-99974574 on Build GRCh38
rs752805401 Chr.15: 99974422-99974422 on Build GRCh38
rs777160312 Chr.15: 99974503-99974503 on Build GRCh38
rs552244295 Chr.15: 99974427-99974427 on Build GRCh38
rs770889652 Chr.15: 99974533-99974533 on Build GRCh38
rs772111759 Chr.15: 99974478-99974478 on Build GRCh38
rs78682885 Chr.15: 99974471-99974471 on Build GRCh38
rs934150528 Chr.15: 99974403-99974403 on Build GRCh38
rs756484586 Chr.15: 99974396-99974396 on Build GRCh38
rs767614213 Chr.15: 99974436-99974436 on Build GRCh38
rs775402406 Chr.15: 99974451-99974451 on Build GRCh38
rs745497667 Chr.15: 99974514-99974514 on Build GRCh38
rs778582648 Chr.15: 99974406-99974406 on Build GRCh38
rs201896149 Chr.15: 99974415-99974415 on Build GRCh38
rs767596205 Chr.15: 99974387-99974387 on Build GRCh38
rs2573652 Chr.15: 99974409-99974409 on Build GRCh38
rs750652557 Chr.15: 99974438-99974438 on Build GRCh38
rs763372199 Chr.15: 99974542-99974542 on Build GRCh38
rs780775353 Chr.15: 99974512-99974512 on Build GRCh38
rs753229004 Chr.15: 99974452-99974452 on Build GRCh38
rs747317920 Chr.15: 99974538-99974538 on Build GRCh38
rs760023197 Chr.15: 99974488-99974488 on Build GRCh38
rs759480599 Chr.15: 99974424-99974424 on Build GRCh38
rs748780450 Chr.15: 99974498-99974499 on Build GRCh38
rs777496319 Chr.15: 99974389-99974389 on Build GRCh38
rs758924018 Chr.15: 99974498-99974498 on Build GRCh38
rs749048665 Chr.15: 99974579-99974579 on Build GRCh38
rs376675165 Chr.15: 99974483-99974483 on Build GRCh38
rs141373128 Chr.15: 99974525-99974525 on Build GRCh38
rs753320297 Chr.15: 99974494-99974494 on Build GRCh38
rs756890553 Chr.15: 99974506-99974506 on Build GRCh38
rs369831239 Chr.15: 99974388-99974388 on Build GRCh38
rs757883707 Chr.15: 99974453-99974453 on Build GRCh38
rs144817994 Chr.15: 99974405-99974405 on Build GRCh38
rs201874119 Chr.15: 99974477-99974477 on Build GRCh38
rs376423645 Chr.15: 99974505-99974505 on Build GRCh38
rs749245963 Chr.15: 99974519-99974519 on Build GRCh38
rs113885511 Chr.15: 99974578-99974578 on Build GRCh38
rs1038775149 Chr.15: 99974411-99974411 on Build GRCh38
rs373278935 Chr.15: 99974431-99974431 on Build GRCh38
rs138405311 Chr.15: 99974487-99974487 on Build GRCh38

View Full Product Details