Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139548286-139548559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F9
Entrez Gene ID
2158
Gene Name
coagulation factor IX
Gene Aliases
F9 p22, FIX, HEMB, P19, PTC, THPH8
UniGene
Hs.522798
Gene Chromosome Location
Chr.X: 139530720-139563459 on Build GRCh38
Transcripts
NM_000133.3 NM_001313913.1 XM_005262397.4

SNP Detail

SNP ID Chromosome Location
rs779281053 Chr.X: 139548456-139548456 on Build GRCh38
rs769388791 Chr.X: 139548341-139548344 on Build GRCh38
rs753711944 Chr.X: 139548321-139548321 on Build GRCh38
rs750611141 Chr.X: 139548395-139548395 on Build GRCh38
rs1047289457 Chr.X: 139548454-139548454 on Build GRCh38
rs984366952 Chr.X: 139548471-139548471 on Build GRCh38
rs137852272 Chr.X: 139548455-139548455 on Build GRCh38
rs780347965 Chr.X: 139548427-139548427 on Build GRCh38
rs865782271 Chr.X: 139548488-139548488 on Build GRCh38
rs45473300 Chr.X: 139548484-139548484 on Build GRCh38
rs765091341 Chr.X: 139548343-139548343 on Build GRCh38
rs759460857 Chr.X: 139548329-139548329 on Build GRCh38
rs767484990 Chr.X: 139548448-139548448 on Build GRCh38
rs1039672013 Chr.X: 139548336-139548336 on Build GRCh38
rs188643991 Chr.X: 139548507-139548507 on Build GRCh38
rs375658633 Chr.X: 139548436-139548436 on Build GRCh38
rs144314232 Chr.X: 139548430-139548430 on Build GRCh38
rs137852236 Chr.X: 139548467-139548467 on Build GRCh38
rs756358425 Chr.X: 139548422-139548422 on Build GRCh38
rs776502394 Chr.X: 139548515-139548515 on Build GRCh38
rs762203336 Chr.X: 139548519-139548520 on Build GRCh38
rs757272947 Chr.X: 139548356-139548356 on Build GRCh38
rs565055999 Chr.X: 139548478-139548478 on Build GRCh38
rs961830711 Chr.X: 139548320-139548320 on Build GRCh38
rs104894807 Chr.X: 139548404-139548404 on Build GRCh38
rs368644284 Chr.X: 139548510-139548510 on Build GRCh38
rs772883788 Chr.X: 139548499-139548499 on Build GRCh38
rs150041781 Chr.X: 139548394-139548394 on Build GRCh38
rs137852235 Chr.X: 139548450-139548450 on Build GRCh38
rs752747648 Chr.X: 139548347-139548347 on Build GRCh38
rs776499043 Chr.X: 139548508-139548508 on Build GRCh38
rs1027316695 Chr.X: 139548353-139548353 on Build GRCh38

View Full Product Details