Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144450674-144450929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYHR1
Entrez Gene ID
50626
Gene Name
cysteine and histidine rich 1
Gene Aliases
CHRP
UniGene
Hs.459379
Gene Chromosome Location
Chr.8: 144449582-144465692 on Build GRCh38
Transcripts
NM_138496.1 XM_006716562.3 XM_011517051.1 XM_017013463.1 XM_017013464.1 XM_017013465.1 XM_017013466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4773098 c.786G>T p.W262C Chr.8: 144450682-144450682 on Build GRCh38
7300482 c.737C>T p.T246M Chr.8: 144450731-144450731 on Build GRCh38
5988139 c.710G>A p.R237Q Chr.8: 144450758-144450758 on Build GRCh38
6516855 c.780G>T p.Q260H Chr.8: 144450688-144450688 on Build GRCh38
4420139 c.733A>C p.I245L Chr.8: 144450735-144450735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868267233 Chr.8: 144450824-144450824 on Build GRCh38
rs774652958 Chr.8: 144450777-144450780 on Build GRCh38
rs908981873 Chr.8: 144450894-144450894 on Build GRCh38
rs759526224 Chr.8: 144450791-144450791 on Build GRCh38
rs964506337 Chr.8: 144450869-144450869 on Build GRCh38
rs917345596 Chr.8: 144450851-144450851 on Build GRCh38
rs905779871 Chr.8: 144450866-144450866 on Build GRCh38
rs540144156 Chr.8: 144450754-144450754 on Build GRCh38
rs577942508 Chr.8: 144450895-144450895 on Build GRCh38
rs771879806 Chr.8: 144450720-144450720 on Build GRCh38
rs909894076 Chr.8: 144450832-144450832 on Build GRCh38
rs949902524 Chr.8: 144450864-144450864 on Build GRCh38
rs770295065 Chr.8: 144450742-144450742 on Build GRCh38
rs556846797 Chr.8: 144450888-144450888 on Build GRCh38
rs774513270 Chr.8: 144450819-144450819 on Build GRCh38
rs770912811 Chr.8: 144450784-144450784 on Build GRCh38
rs747702632 Chr.8: 144450712-144450712 on Build GRCh38
rs538277145 Chr.8: 144450880-144450880 on Build GRCh38
rs1017357362 Chr.8: 144450867-144450867 on Build GRCh38
rs1044655877 Chr.8: 144450865-144450865 on Build GRCh38
rs1030185058 Chr.8: 144450887-144450887 on Build GRCh38
rs566965865 Chr.8: 144450709-144450709 on Build GRCh38
rs75215796 Chr.8: 144450844-144450844 on Build GRCh38
rs746781715 Chr.8: 144450768-144450768 on Build GRCh38
rs1002425442 Chr.8: 144450792-144450792 on Build GRCh38
rs905447693 Chr.8: 144450757-144450757 on Build GRCh38
rs1054090977 Chr.8: 144450892-144450892 on Build GRCh38
rs549701114 Chr.8: 144450807-144450807 on Build GRCh38
rs773453591 Chr.8: 144450850-144450850 on Build GRCh38
rs143645257 Chr.8: 144450800-144450800 on Build GRCh38
rs921516679 Chr.8: 144450802-144450802 on Build GRCh38
rs983492730 Chr.8: 144450893-144450893 on Build GRCh38
rs776430893 Chr.8: 144450786-144450786 on Build GRCh38

View Full Product Details