Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75600165-75600430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAT1
Entrez Gene ID
23536
Gene Name
adenosine deaminase, tRNA specific 1
Gene Aliases
HADAT1
UniGene
Hs.729312
Gene Chromosome Location
Chr.16: 75596879-75623323 on Build GRCh38
Transcripts
NM_001324444.1 NM_001324445.1 NM_001324446.1 NM_001324448.1 NM_001324449.1 NM_001324450.1 NM_001324451.1 NM_001324452.1 NM_001324453.1 NM_012091.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4413387 c.1428C>A p.Y476* Chr.16: 75600297-75600297 on Build GRCh38
349243 c.1473G>T p.W491C Chr.16: 75600252-75600252 on Build GRCh38
272314 c.1418C>T p.A473V Chr.16: 75600307-75600307 on Build GRCh38
331821 c.1444A>G p.T482A Chr.16: 75600281-75600281 on Build GRCh38
1379817 c.1438T>C p.W480R Chr.16: 75600287-75600287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768358891 Chr.16: 75600238-75600238 on Build GRCh38
rs754878846 Chr.16: 75600307-75600307 on Build GRCh38
rs750044883 Chr.16: 75600285-75600285 on Build GRCh38
rs371126400 Chr.16: 75600241-75600241 on Build GRCh38
rs763117593 Chr.16: 75600347-75600347 on Build GRCh38
rs996127329 Chr.16: 75600330-75600330 on Build GRCh38
rs765710834 Chr.16: 75600248-75600248 on Build GRCh38
rs751180473 Chr.16: 75600188-75600188 on Build GRCh38
rs75248264 Chr.16: 75600294-75600294 on Build GRCh38
rs776176191 Chr.16: 75600260-75600260 on Build GRCh38
rs571304159 Chr.16: 75600397-75600397 on Build GRCh38
rs762276584 Chr.16: 75600384-75600384 on Build GRCh38
rs1045017615 Chr.16: 75600197-75600197 on Build GRCh38
rs780700881 Chr.16: 75600209-75600209 on Build GRCh38
rs776706277 Chr.16: 75600243-75600243 on Build GRCh38
rs752025717 Chr.16: 75600275-75600275 on Build GRCh38
rs192646642 Chr.16: 75600227-75600227 on Build GRCh38
rs367677899 Chr.16: 75600252-75600252 on Build GRCh38
rs757546362 Chr.16: 75600276-75600276 on Build GRCh38
rs764367463 Chr.16: 75600274-75600274 on Build GRCh38
rs764084011 Chr.16: 75600278-75600278 on Build GRCh38
rs780006147 Chr.16: 75600226-75600226 on Build GRCh38
rs775650625 Chr.16: 75600409-75600409 on Build GRCh38
rs777036429 Chr.16: 75600317-75600317 on Build GRCh38
rs769261760 Chr.16: 75600375-75600375 on Build GRCh38
rs376302952 Chr.16: 75600242-75600242 on Build GRCh38
rs746050947 Chr.16: 75600328-75600328 on Build GRCh38
rs750098288 Chr.16: 75600396-75600396 on Build GRCh38
rs188035836 Chr.16: 75600187-75600187 on Build GRCh38
rs771744048 Chr.16: 75600316-75600316 on Build GRCh38
rs778712606 Chr.16: 75600311-75600311 on Build GRCh38
rs868261779 Chr.16: 75600234-75600234 on Build GRCh38
rs745438639 Chr.16: 75600222-75600222 on Build GRCh38
rs763510007 Chr.16: 75600269-75600269 on Build GRCh38
rs199566267 Chr.16: 75600385-75600385 on Build GRCh38
rs200186570 Chr.16: 75600334-75600334 on Build GRCh38
rs765878755 Chr.16: 75600400-75600400 on Build GRCh38
rs947650968 Chr.16: 75600192-75600192 on Build GRCh38
rs749183266 Chr.16: 75600237-75600237 on Build GRCh38
rs774695638 Chr.16: 75600378-75600378 on Build GRCh38
rs1057294764 Chr.16: 75600268-75600268 on Build GRCh38
rs367944178 Chr.16: 75600344-75600344 on Build GRCh38
rs1048401322 Chr.16: 75600196-75600196 on Build GRCh38
rs987748977 Chr.16: 75600394-75600394 on Build GRCh38
rs144876601 Chr.16: 75600297-75600297 on Build GRCh38
rs749236621 Chr.16: 75600306-75600306 on Build GRCh38
rs368333664 Chr.16: 75600189-75600189 on Build GRCh38

View Full Product Details