Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33092385-33092650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFB4
Entrez Gene ID
149954
Gene Name
BPI fold containing family B member 4
Gene Aliases
C20orf186, LPLUNC4, RY2G5, dJ726C3.5
UniGene
Hs.38961
Gene Chromosome Location
Chr.20: 33081512-33111751 on Build GRCh38
Transcripts
NM_182519.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
215810 c.1064A>C p.Y355S Chr.20: 33092495-33092495 on Build GRCh38
350179 c.1164G>T p.L388L Chr.20: 33092595-33092595 on Build GRCh38
6377336 c.1158C>A p.N386K Chr.20: 33092589-33092589 on Build GRCh38
3939304 c.1210G>T p.D404Y Chr.20: 33092641-33092641 on Build GRCh38
3389683 c.1059C>T p.I353I Chr.20: 33092490-33092490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746344980 Chr.20: 33092442-33092442 on Build GRCh38
rs985341265 Chr.20: 33092475-33092475 on Build GRCh38
rs967967015 Chr.20: 33092556-33092556 on Build GRCh38
rs755760238 Chr.20: 33092447-33092448 on Build GRCh38
rs754783912 Chr.20: 33092580-33092580 on Build GRCh38
rs772743997 Chr.20: 33092519-33092519 on Build GRCh38
rs753678971 Chr.20: 33092576-33092576 on Build GRCh38
rs770283793 Chr.20: 33092623-33092623 on Build GRCh38
rs372865308 Chr.20: 33092533-33092533 on Build GRCh38
rs574793283 Chr.20: 33092605-33092605 on Build GRCh38
rs546005446 Chr.20: 33092409-33092409 on Build GRCh38
rs372775889 Chr.20: 33092451-33092451 on Build GRCh38
rs777210954 Chr.20: 33092612-33092612 on Build GRCh38
rs764228705 Chr.20: 33092471-33092471 on Build GRCh38
rs768285033 Chr.20: 33092504-33092504 on Build GRCh38
rs151182059 Chr.20: 33092459-33092459 on Build GRCh38
rs17122711 Chr.20: 33092528-33092528 on Build GRCh38
rs778602410 Chr.20: 33092581-33092581 on Build GRCh38
rs780142226 Chr.20: 33092559-33092559 on Build GRCh38
rs200333820 Chr.20: 33092529-33092529 on Build GRCh38
rs759991327 Chr.20: 33092523-33092523 on Build GRCh38
rs771151395 Chr.20: 33092406-33092406 on Build GRCh38
rs997569885 Chr.20: 33092573-33092573 on Build GRCh38
rs779945261 Chr.20: 33092500-33092500 on Build GRCh38
rs923242407 Chr.20: 33092463-33092463 on Build GRCh38
rs767455154 Chr.20: 33092544-33092544 on Build GRCh38
rs753085448 Chr.20: 33092461-33092461 on Build GRCh38
rs199542008 Chr.20: 33092557-33092557 on Build GRCh38
rs779596543 Chr.20: 33092476-33092478 on Build GRCh38
rs113445311 Chr.20: 33092526-33092526 on Build GRCh38
rs776035940 Chr.20: 33092627-33092627 on Build GRCh38
rs1038188451 Chr.20: 33092491-33092491 on Build GRCh38
rs761921641 Chr.20: 33092539-33092539 on Build GRCh38
rs929714505 Chr.20: 33092497-33092497 on Build GRCh38
rs766902471 Chr.20: 33092456-33092456 on Build GRCh38
rs2070327 Chr.20: 33092428-33092428 on Build GRCh38
rs750552572 Chr.20: 33092545-33092545 on Build GRCh38
rs868818866 Chr.20: 33092594-33092594 on Build GRCh38
rs369095045 Chr.20: 33092477-33092477 on Build GRCh38
rs139946022 Chr.20: 33092482-33092482 on Build GRCh38
rs140173449 Chr.20: 33092473-33092473 on Build GRCh38
rs141724545 Chr.20: 33092492-33092492 on Build GRCh38
rs771527154 Chr.20: 33092518-33092518 on Build GRCh38
rs748907564 Chr.20: 33092418-33092418 on Build GRCh38
rs893368999 Chr.20: 33092532-33092532 on Build GRCh38
rs775251912 Chr.20: 33092415-33092415 on Build GRCh38
rs758561468 Chr.20: 33092465-33092465 on Build GRCh38
rs773850988 Chr.20: 33092450-33092450 on Build GRCh38
rs765549300 Chr.20: 33092460-33092460 on Build GRCh38
rs564724845 Chr.20: 33092505-33092505 on Build GRCh38
rs756055417 Chr.20: 33092546-33092546 on Build GRCh38
rs544445188 Chr.20: 33092503-33092503 on Build GRCh38
rs1011740981 Chr.20: 33092414-33092414 on Build GRCh38
rs747677261 Chr.20: 33092515-33092515 on Build GRCh38
rs147935083 Chr.20: 33092490-33092490 on Build GRCh38
rs768941830 Chr.20: 33092611-33092612 on Build GRCh38
rs774901152 Chr.20: 33092614-33092615 on Build GRCh38
rs746529134 Chr.20: 33092620-33092620 on Build GRCh38
rs756150390 Chr.20: 33092496-33092496 on Build GRCh38
rs890778791 Chr.20: 33092571-33092571 on Build GRCh38
rs747720586 Chr.20: 33092589-33092589 on Build GRCh38
rs749627205 Chr.20: 33092491-33092491 on Build GRCh38

View Full Product Details