Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121835235-121835506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAF2
Entrez Gene ID
55840
Gene Name
ELL associated factor 2
Gene Aliases
BM040, TRAITS, U19
UniGene
Hs.477325
Gene Chromosome Location
Chr.3: 121835180-121886526 on Build GRCh38
Transcripts
NM_001320041.1 NM_018456.5 XM_005247618.3 XM_017006861.1 XM_017006863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3408169 c.9C>A p.S3R Chr.3: 121835294-121835294 on Build GRCh38
4157172 c.82C>G p.R28G Chr.3: 121835367-121835367 on Build GRCh38
1308461 c.21C>T p.F7F Chr.3: 121835306-121835306 on Build GRCh38
3940229 c.31G>A p.D11N Chr.3: 121835316-121835316 on Build GRCh38
3586049 c.24A>T p.S8S Chr.3: 121835309-121835309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920514260 Chr.3: 121835416-121835416 on Build GRCh38
rs775341123 Chr.3: 121835318-121835318 on Build GRCh38
rs753873255 Chr.3: 121835322-121835322 on Build GRCh38
rs528607299 Chr.3: 121835291-121835291 on Build GRCh38
rs755018682 Chr.3: 121835323-121835323 on Build GRCh38
rs371705531 Chr.3: 121835408-121835408 on Build GRCh38
rs570523602 Chr.3: 121835328-121835328 on Build GRCh38
rs757640630 Chr.3: 121835436-121835436 on Build GRCh38
rs13098802 Chr.3: 121835426-121835426 on Build GRCh38
rs201683243 Chr.3: 121835255-121835255 on Build GRCh38
rs113829843 Chr.3: 121835300-121835300 on Build GRCh38
rs368458597 Chr.3: 121835327-121835327 on Build GRCh38
rs751927784 Chr.3: 121835423-121835423 on Build GRCh38
rs778143626 Chr.3: 121835350-121835350 on Build GRCh38
rs761640397 Chr.3: 121835404-121835404 on Build GRCh38
rs910272981 Chr.3: 121835438-121835438 on Build GRCh38
rs780831957 Chr.3: 121835372-121835372 on Build GRCh38
rs745458204 Chr.3: 121835394-121835395 on Build GRCh38
rs766569686 Chr.3: 121835302-121835302 on Build GRCh38
rs151186324 Chr.3: 121835362-121835362 on Build GRCh38
rs765337013 Chr.3: 121835415-121835415 on Build GRCh38
rs769626878 Chr.3: 121835263-121835263 on Build GRCh38
rs937925819 Chr.3: 121835320-121835320 on Build GRCh38
rs768672221 Chr.3: 121835391-121835391 on Build GRCh38
rs1040549085 Chr.3: 121835457-121835457 on Build GRCh38
rs944444682 Chr.3: 121835456-121835456 on Build GRCh38
rs749210328 Chr.3: 121835388-121835388 on Build GRCh38
rs750711954 Chr.3: 121835294-121835294 on Build GRCh38
rs780768241 Chr.3: 121835439-121835439 on Build GRCh38
rs747327741 Chr.3: 121835361-121835361 on Build GRCh38
rs1002382852 Chr.3: 121835321-121835321 on Build GRCh38
rs757935217 Chr.3: 121835431-121835434 on Build GRCh38
rs774307699 Chr.3: 121835398-121835398 on Build GRCh38
rs776028421 Chr.3: 121835253-121835253 on Build GRCh38
rs758810086 Chr.3: 121835348-121835348 on Build GRCh38
rs758927625 Chr.3: 121835420-121835420 on Build GRCh38
rs752735565 Chr.3: 121835418-121835418 on Build GRCh38
rs556452858 Chr.3: 121835380-121835380 on Build GRCh38
rs762901645 Chr.3: 121835289-121835289 on Build GRCh38
rs985945519 Chr.3: 121835434-121835434 on Build GRCh38
rs745440983 Chr.3: 121835373-121835373 on Build GRCh38
rs535424869 Chr.3: 121835453-121835453 on Build GRCh38
rs954302405 Chr.3: 121835440-121835440 on Build GRCh38
rs930438671 Chr.3: 121835433-121835433 on Build GRCh38
rs201253070 Chr.3: 121835407-121835407 on Build GRCh38
rs775496917 Chr.3: 121835278-121835278 on Build GRCh38
rs775341753 Chr.3: 121835382-121835382 on Build GRCh38
rs1054741789 Chr.3: 121835324-121835324 on Build GRCh38
rs3805125 Chr.3: 121835482-121835482 on Build GRCh38

View Full Product Details