Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96263641-96263902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDKRB1
Entrez Gene ID
623
Gene Name
bradykinin receptor B1
Gene Aliases
B1BKR, B1R, BDKRB2, BKB1R, BKR1, BRADYB1
UniGene
Hs.525572
Gene Chromosome Location
Chr.14: 96256210-96264763 on Build GRCh38
Transcripts
NM_000710.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4578089 c.197G>A p.R66Q Chr.14: 96263879-96263879 on Build GRCh38
341714 c.110A>G p.H37R Chr.14: 96263792-96263792 on Build GRCh38
5504075 c.15G>A p.W5* Chr.14: 96263697-96263697 on Build GRCh38
5971316 c.11C>T p.S4F Chr.14: 96263693-96263693 on Build GRCh38
376629 c.195C>A p.P65P Chr.14: 96263877-96263877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139498601 Chr.14: 96263710-96263710 on Build GRCh38
rs145437913 Chr.14: 96263754-96263754 on Build GRCh38
rs1000091722 Chr.14: 96263797-96263797 on Build GRCh38
rs766803564 Chr.14: 96263883-96263883 on Build GRCh38
rs769845038 Chr.14: 96263848-96263848 on Build GRCh38
rs773146412 Chr.14: 96263856-96263856 on Build GRCh38
rs755158731 Chr.14: 96263819-96263819 on Build GRCh38
rs762659699 Chr.14: 96263708-96263708 on Build GRCh38
rs149672766 Chr.14: 96263737-96263737 on Build GRCh38
rs775407674 Chr.14: 96263769-96263769 on Build GRCh38
rs757769026 Chr.14: 96263725-96263725 on Build GRCh38
rs201013948 Chr.14: 96263872-96263872 on Build GRCh38
rs200329555 Chr.14: 96263777-96263777 on Build GRCh38
rs779136878 Chr.14: 96263823-96263823 on Build GRCh38
rs200706096 Chr.14: 96263882-96263882 on Build GRCh38
rs148906103 Chr.14: 96263783-96263783 on Build GRCh38
rs369051092 Chr.14: 96263686-96263686 on Build GRCh38
rs147639735 Chr.14: 96263804-96263804 on Build GRCh38
rs762755879 Chr.14: 96263770-96263770 on Build GRCh38
rs774007342 Chr.14: 96263878-96263878 on Build GRCh38
rs372076696 Chr.14: 96263735-96263735 on Build GRCh38
rs372693065 Chr.14: 96263793-96263793 on Build GRCh38
rs145413716 Chr.14: 96263832-96263832 on Build GRCh38
rs765008189 Chr.14: 96263693-96263693 on Build GRCh38
rs761261697 Chr.14: 96263879-96263879 on Build GRCh38
rs768352660 Chr.14: 96263771-96263771 on Build GRCh38
rs771652663 Chr.14: 96263880-96263880 on Build GRCh38
rs571018823 Chr.14: 96263757-96263757 on Build GRCh38
rs775087789 Chr.14: 96263702-96263702 on Build GRCh38
rs769960015 Chr.14: 96263678-96263679 on Build GRCh38
rs201572929 Chr.14: 96263845-96263845 on Build GRCh38
rs369936721 Chr.14: 96263833-96263833 on Build GRCh38
rs145317133 Chr.14: 96263802-96263802 on Build GRCh38
rs369133566 Chr.14: 96263745-96263745 on Build GRCh38
rs2071084 Chr.14: 96263805-96263805 on Build GRCh38
rs373000921 Chr.14: 96263881-96263881 on Build GRCh38
rs778928912 Chr.14: 96263753-96263753 on Build GRCh38
rs938159640 Chr.14: 96263673-96263673 on Build GRCh38
rs886632520 Chr.14: 96263681-96263681 on Build GRCh38
rs780064132 Chr.14: 96263849-96263849 on Build GRCh38
rs768209755 Chr.14: 96263871-96263871 on Build GRCh38
rs528094974 Chr.14: 96263728-96263728 on Build GRCh38
rs376961698 Chr.14: 96263744-96263744 on Build GRCh38
rs769747045 Chr.14: 96263767-96263767 on Build GRCh38
rs745685069 Chr.14: 96263766-96263766 on Build GRCh38
rs377450315 Chr.14: 96263800-96263800 on Build GRCh38
rs751634375 Chr.14: 96263845-96263853 on Build GRCh38
rs745854535 Chr.14: 96263847-96263847 on Build GRCh38
rs201942990 Chr.14: 96263715-96263715 on Build GRCh38
rs749027831 Chr.14: 96263861-96263861 on Build GRCh38
rs776296998 Chr.14: 96263676-96263676 on Build GRCh38

View Full Product Details