Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134741139-134741406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMLG
Entrez Gene ID
819
Gene Name
calcium modulating ligand
Gene Aliases
CAML, GET2
UniGene
Hs.529846
Gene Chromosome Location
Chr.5: 134737927-134752160 on Build GRCh38
Transcripts
NM_001745.3 XM_011543653.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3850203 c.446G>A p.R149H Chr.5: 134741336-134741336 on Build GRCh38
1433191 c.250C>T p.R84* Chr.5: 134741140-134741140 on Build GRCh38
6339079 c.289G>A p.D97N Chr.5: 134741179-134741179 on Build GRCh38
4854890 c.310G>A p.E104K Chr.5: 134741200-134741200 on Build GRCh38
3850202 c.423G>A p.A141A Chr.5: 134741313-134741313 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769734815 Chr.5: 134741176-134741176 on Build GRCh38
rs138877680 Chr.5: 134741335-134741335 on Build GRCh38
rs968265332 Chr.5: 134741230-134741230 on Build GRCh38
rs200289909 Chr.5: 134741234-134741234 on Build GRCh38
rs781423962 Chr.5: 134741338-134741338 on Build GRCh38
rs188626944 Chr.5: 134741285-134741285 on Build GRCh38
rs773877145 Chr.5: 134741291-134741291 on Build GRCh38
rs775159638 Chr.5: 134741283-134741283 on Build GRCh38
rs143564828 Chr.5: 134741200-134741200 on Build GRCh38
rs143592213 Chr.5: 134741273-134741273 on Build GRCh38
rs757373789 Chr.5: 134741336-134741336 on Build GRCh38
rs765623837 Chr.5: 134741317-134741317 on Build GRCh38
rs756063078 Chr.5: 134741367-134741367 on Build GRCh38
rs569294650 Chr.5: 134741318-134741318 on Build GRCh38
rs777907734 Chr.5: 134741248-134741248 on Build GRCh38
rs768329760 Chr.5: 134741375-134741375 on Build GRCh38
rs774383139 Chr.5: 134741282-134741283 on Build GRCh38
rs769564299 Chr.5: 134741276-134741276 on Build GRCh38
rs185311575 Chr.5: 134741232-134741232 on Build GRCh38
rs763600778 Chr.5: 134741194-134741194 on Build GRCh38
rs777080259 Chr.5: 134741304-134741304 on Build GRCh38
rs776535781 Chr.5: 134741169-134741169 on Build GRCh38
rs757239147 Chr.5: 134741257-134741257 on Build GRCh38
rs754177604 Chr.5: 134741202-134741202 on Build GRCh38
rs775167047 Chr.5: 134741184-134741184 on Build GRCh38
rs11552197 Chr.5: 134741188-134741188 on Build GRCh38
rs768511656 Chr.5: 134741266-134741267 on Build GRCh38
rs138133729 Chr.5: 134741268-134741268 on Build GRCh38
rs373460413 Chr.5: 134741275-134741275 on Build GRCh38
rs759454520 Chr.5: 134741172-134741172 on Build GRCh38
rs745740360 Chr.5: 134741342-134741342 on Build GRCh38
rs201890510 Chr.5: 134741161-134741161 on Build GRCh38
rs201335217 Chr.5: 134741312-134741312 on Build GRCh38
rs752908968 Chr.5: 134741235-134741235 on Build GRCh38
rs528276022 Chr.5: 134741199-134741199 on Build GRCh38
rs1014541383 Chr.5: 134741314-134741314 on Build GRCh38
rs755283193 Chr.5: 134741218-134741218 on Build GRCh38
rs758619095 Chr.5: 134741319-134741319 on Build GRCh38
rs370493122 Chr.5: 134741238-134741238 on Build GRCh38
rs772043905 Chr.5: 134741371-134741371 on Build GRCh38
rs371384638 Chr.5: 134741355-134741355 on Build GRCh38
rs1015570441 Chr.5: 134741289-134741289 on Build GRCh38
rs146174222 Chr.5: 134741284-134741284 on Build GRCh38
rs558413025 Chr.5: 134741374-134741374 on Build GRCh38
rs374260960 Chr.5: 134741292-134741292 on Build GRCh38
rs1028218740 Chr.5: 134741266-134741266 on Build GRCh38
rs767032715 Chr.5: 134741295-134741295 on Build GRCh38
rs773941201 Chr.5: 134741377-134741377 on Build GRCh38
rs747126920 Chr.5: 134741252-134741252 on Build GRCh38
rs377383964 Chr.5: 134741333-134741333 on Build GRCh38
rs11552198 Chr.5: 134741212-134741212 on Build GRCh38

View Full Product Details