Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 100007499-100007760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERP44
Entrez Gene ID
23071
Gene Name
endoplasmic reticulum protein 44
Gene Aliases
PDIA10, TXNDC4
UniGene
Hs.154023
Gene Chromosome Location
Chr.9: 99979179-100099052 on Build GRCh38
Transcripts
NM_015051.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5648396 c.872A>C p.K291T Chr.9: 100007580-100007580 on Build GRCh38
6367445 c.863T>A p.I288K Chr.9: 100007589-100007589 on Build GRCh38
6656268 c.873delA p.G292fs*3 Chr.9: 100007579-100007579 on Build GRCh38
6182229 c.785C>T p.P262L Chr.9: 100007667-100007667 on Build GRCh38
7111371 c.820G>C p.E274Q Chr.9: 100007632-100007632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964566304 Chr.9: 100007695-100007695 on Build GRCh38
rs757764057 Chr.9: 100007594-100007594 on Build GRCh38
rs761829637 Chr.9: 100007533-100007533 on Build GRCh38
rs187892197 Chr.9: 100007535-100007535 on Build GRCh38
rs779325291 Chr.9: 100007530-100007530 on Build GRCh38
rs778224200 Chr.9: 100007576-100007576 on Build GRCh38
rs377604852 Chr.9: 100007727-100007727 on Build GRCh38
rs767904386 Chr.9: 100007551-100007551 on Build GRCh38
rs1020643733 Chr.9: 100007540-100007540 on Build GRCh38
rs756251376 Chr.9: 100007564-100007564 on Build GRCh38
rs368620789 Chr.9: 100007543-100007543 on Build GRCh38
rs879219513 Chr.9: 100007730-100007730 on Build GRCh38
rs745440655 Chr.9: 100007628-100007628 on Build GRCh38
rs371347560 Chr.9: 100007636-100007636 on Build GRCh38
rs779387092 Chr.9: 100007601-100007601 on Build GRCh38
rs878946630 Chr.9: 100007694-100007694 on Build GRCh38
rs879015161 Chr.9: 100007692-100007692 on Build GRCh38
rs752933438 Chr.9: 100007558-100007558 on Build GRCh38
rs765202497 Chr.9: 100007534-100007534 on Build GRCh38
rs776491816 Chr.9: 100007687-100007687 on Build GRCh38
rs113208535 Chr.9: 100007684-100007684 on Build GRCh38
rs761511347 Chr.9: 100007704-100007704 on Build GRCh38
rs768142376 Chr.9: 100007686-100007686 on Build GRCh38
rs761880029 Chr.9: 100007538-100007538 on Build GRCh38
rs779608928 Chr.9: 100007660-100007660 on Build GRCh38
rs754095141 Chr.9: 100007589-100007589 on Build GRCh38

View Full Product Details