Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 26211603-26211842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX3
Entrez Gene ID
11335
Gene Name
chromobox 3
Gene Aliases
HECH, HP1-GAMMA, HP1Hs-gamma
UniGene
Hs.381189
Gene Chromosome Location
Chr.7: 26201211-26213607 on Build GRCh38
Transcripts
NM_007276.4 NM_016587.3 XM_005249611.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
452905 c.420G>A p.M140I Chr.7: 26211751-26211751 on Build GRCh38
5407279 c.414T>C p.F138F Chr.7: 26211745-26211745 on Build GRCh38
7168855 c.370G>A p.E124K Chr.7: 26211701-26211701 on Build GRCh38
5438206 c.367C>T p.P123S Chr.7: 26211698-26211698 on Build GRCh38
4598959 c.411G>A p.M137I Chr.7: 26211742-26211742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950375819 Chr.7: 26211641-26211642 on Build GRCh38
rs765839751 Chr.7: 26211774-26211774 on Build GRCh38
rs773462431 Chr.7: 26211798-26211798 on Build GRCh38
rs745745464 Chr.7: 26211663-26211663 on Build GRCh38
rs768288217 Chr.7: 26211741-26211741 on Build GRCh38
rs771247002 Chr.7: 26211761-26211761 on Build GRCh38
rs759917188 Chr.7: 26211765-26211765 on Build GRCh38
rs771008947 Chr.7: 26211754-26211754 on Build GRCh38
rs376630956 Chr.7: 26211685-26211685 on Build GRCh38
rs759293919 Chr.7: 26211797-26211798 on Build GRCh38
rs769756418 Chr.7: 26211666-26211666 on Build GRCh38
rs76937019 Chr.7: 26211755-26212080 on Build GRCh38
rs372921828 Chr.7: 26211763-26211763 on Build GRCh38
rs368955032 Chr.7: 26211803-26211803 on Build GRCh38
rs748257690 Chr.7: 26211767-26211768 on Build GRCh38
rs764405458 Chr.7: 26211808-26211808 on Build GRCh38
rs200791010 Chr.7: 26211748-26211748 on Build GRCh38
rs921290504 Chr.7: 26211651-26211651 on Build GRCh38
rs374663838 Chr.7: 26211778-26211778 on Build GRCh38
rs200410047 Chr.7: 26211718-26211718 on Build GRCh38
rs774507846 Chr.7: 26211764-26211765 on Build GRCh38
rs772240792 Chr.7: 26211794-26211804 on Build GRCh38
rs905899027 Chr.7: 26211773-26211773 on Build GRCh38
rs768878661 Chr.7: 26211659-26211660 on Build GRCh38
rs775430990 Chr.7: 26211798-26211802 on Build GRCh38
rs764982161 Chr.7: 26211798-26211799 on Build GRCh38
rs543343409 Chr.7: 26211809-26211809 on Build GRCh38
rs749771730 Chr.7: 26211640-26211649 on Build GRCh38

View Full Product Details