Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32415784-32416057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFC
Entrez Gene ID
254240
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
UniGene
Hs.372939
Gene Chromosome Location
Chr.22: 32413845-32464465 on Build GRCh38
Transcripts
NM_174932.2 XM_011530088.2 XM_011530089.1 XM_011530090.1 XM_011530091.2 XM_017028740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
444914 c.1351T>C p.S451P Chr.22: 32415965-32415965 on Build GRCh38
274152 c.1341A>G p.G447G Chr.22: 32415975-32415975 on Build GRCh38
7149552 c.1399G>C p.E467Q Chr.22: 32415917-32415917 on Build GRCh38
4526364 c.1390G>A p.E464K Chr.22: 32415926-32415926 on Build GRCh38
5049237 c.1384G>C p.D462H Chr.22: 32415932-32415932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747336667 Chr.22: 32415910-32415910 on Build GRCh38
rs940941338 Chr.22: 32415845-32415845 on Build GRCh38
rs771394597 Chr.22: 32415913-32415913 on Build GRCh38
rs1018646171 Chr.22: 32416031-32416031 on Build GRCh38
rs1031296476 Chr.22: 32415815-32415815 on Build GRCh38
rs757147994 Chr.22: 32415870-32415870 on Build GRCh38
rs750351154 Chr.22: 32416010-32416010 on Build GRCh38
rs376019063 Chr.22: 32415872-32415872 on Build GRCh38
rs893682926 Chr.22: 32415969-32415969 on Build GRCh38
rs911532945 Chr.22: 32415833-32415833 on Build GRCh38
rs779740946 Chr.22: 32416015-32416015 on Build GRCh38
rs903166567 Chr.22: 32415947-32415947 on Build GRCh38
rs374283701 Chr.22: 32416017-32416017 on Build GRCh38
rs373924654 Chr.22: 32415867-32415867 on Build GRCh38
rs773712960 Chr.22: 32415904-32415904 on Build GRCh38
rs778610853 Chr.22: 32416021-32416021 on Build GRCh38
rs754688262 Chr.22: 32416020-32416020 on Build GRCh38
rs200517481 Chr.22: 32415869-32415869 on Build GRCh38
rs576559991 Chr.22: 32415941-32415941 on Build GRCh38
rs960979201 Chr.22: 32415822-32415823 on Build GRCh38
rs867248388 Chr.22: 32415877-32415877 on Build GRCh38
rs768380588 Chr.22: 32415900-32415900 on Build GRCh38
rs779900508 Chr.22: 32415891-32415891 on Build GRCh38
rs201615090 Chr.22: 32415917-32415917 on Build GRCh38
rs992786945 Chr.22: 32415853-32415853 on Build GRCh38
rs745436883 Chr.22: 32415882-32415882 on Build GRCh38
rs761655827 Chr.22: 32415983-32415983 on Build GRCh38
rs115332147 Chr.22: 32415986-32415986 on Build GRCh38
rs5998478 Chr.22: 32415965-32415965 on Build GRCh38
rs751325586 Chr.22: 32415977-32415977 on Build GRCh38
rs769844716 Chr.22: 32415944-32415944 on Build GRCh38
rs565424741 Chr.22: 32415974-32415974 on Build GRCh38
rs1030861095 Chr.22: 32415967-32415967 on Build GRCh38
rs16990421 Chr.22: 32415897-32415897 on Build GRCh38
rs530036846 Chr.22: 32416024-32416024 on Build GRCh38
rs201389531 Chr.22: 32416020-32416038 on Build GRCh38
rs1046159436 Chr.22: 32415938-32415938 on Build GRCh38
rs56965504 Chr.22: 32416011-32416011 on Build GRCh38
rs907833288 Chr.22: 32415984-32415984 on Build GRCh38
rs560915048 Chr.22: 32416019-32416019 on Build GRCh38
rs145148624 Chr.22: 32415973-32415973 on Build GRCh38
rs894553437 Chr.22: 32415911-32415911 on Build GRCh38

View Full Product Details