Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59192047-59192320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTX4
Entrez Gene ID
23220
Gene Name
deltex E3 ubiquitin ligase 4
Gene Aliases
RNF155
UniGene
Hs.523696
Gene Chromosome Location
Chr.11: 59171430-59208587 on Build GRCh38
Transcripts
NM_001300727.1 NM_015177.1 XM_006718483.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6032587 c.934G>A p.A312T Chr.11: 59192128-59192128 on Build GRCh38
5765593 c.1309C>G p.L437V Chr.11: 59192185-59192185 on Build GRCh38
266430 c.911C>T p.T304I Chr.11: 59192105-59192105 on Build GRCh38
4525046 c.986G>T p.G329V Chr.11: 59192180-59192180 on Build GRCh38
6650105 c.1319G>A p.C440Y Chr.11: 59192195-59192195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759665079 Chr.11: 59192256-59192256 on Build GRCh38
rs970303694 Chr.11: 59192118-59192118 on Build GRCh38
rs201305796 Chr.11: 59192119-59192119 on Build GRCh38
rs369979555 Chr.11: 59192088-59192088 on Build GRCh38
rs372596030 Chr.11: 59192150-59192150 on Build GRCh38
rs372183643 Chr.11: 59192066-59192066 on Build GRCh38
rs777456811 Chr.11: 59192111-59192111 on Build GRCh38
rs748661573 Chr.11: 59192212-59192212 on Build GRCh38
rs201910679 Chr.11: 59192159-59192159 on Build GRCh38
rs774800473 Chr.11: 59192282-59192283 on Build GRCh38
rs781709096 Chr.11: 59192204-59192204 on Build GRCh38
rs747460993 Chr.11: 59192121-59192121 on Build GRCh38
rs559768787 Chr.11: 59192151-59192151 on Build GRCh38
rs749850400 Chr.11: 59192175-59192175 on Build GRCh38
rs767370285 Chr.11: 59192083-59192083 on Build GRCh38
rs546051013 Chr.11: 59192197-59192197 on Build GRCh38
rs113471609 Chr.11: 59192296-59192296 on Build GRCh38
rs780461764 Chr.11: 59192151-59192153 on Build GRCh38
rs373376556 Chr.11: 59192192-59192192 on Build GRCh38
rs140997264 Chr.11: 59192294-59192294 on Build GRCh38
rs769175775 Chr.11: 59192126-59192126 on Build GRCh38
rs369820960 Chr.11: 59192074-59192074 on Build GRCh38
rs748517603 Chr.11: 59192130-59192130 on Build GRCh38
rs151058472 Chr.11: 59192244-59192244 on Build GRCh38
rs894854954 Chr.11: 59192070-59192070 on Build GRCh38
rs774593866 Chr.11: 59192153-59192153 on Build GRCh38
rs778348066 Chr.11: 59192198-59192198 on Build GRCh38
rs771307637 Chr.11: 59192246-59192246 on Build GRCh38
rs375172205 Chr.11: 59192067-59192067 on Build GRCh38
rs769205660 Chr.11: 59192276-59192276 on Build GRCh38
rs763335943 Chr.11: 59192148-59192148 on Build GRCh38
rs181187928 Chr.11: 59192079-59192079 on Build GRCh38
rs767209563 Chr.11: 59192139-59192139 on Build GRCh38
rs774753445 Chr.11: 59192247-59192247 on Build GRCh38
rs376347977 Chr.11: 59192235-59192235 on Build GRCh38
rs989617491 Chr.11: 59192142-59192142 on Build GRCh38
rs770192858 Chr.11: 59192228-59192228 on Build GRCh38
rs369788862 Chr.11: 59192147-59192147 on Build GRCh38
rs767458436 Chr.11: 59192155-59192155 on Build GRCh38
rs775488761 Chr.11: 59192266-59192266 on Build GRCh38
rs755901041 Chr.11: 59192106-59192106 on Build GRCh38
rs1025103416 Chr.11: 59192105-59192105 on Build GRCh38
rs753418892 Chr.11: 59192196-59192196 on Build GRCh38
rs770106528 Chr.11: 59192135-59192135 on Build GRCh38
rs759505671 Chr.11: 59192154-59192154 on Build GRCh38
rs375535068 Chr.11: 59192120-59192120 on Build GRCh38
rs749971231 Chr.11: 59192202-59192202 on Build GRCh38
rs761417674 Chr.11: 59192292-59192292 on Build GRCh38
rs376329930 Chr.11: 59192273-59192280 on Build GRCh38
rs757947767 Chr.11: 59192203-59192203 on Build GRCh38
rs763889608 Chr.11: 59192290-59192290 on Build GRCh38
rs1051435553 Chr.11: 59192189-59192189 on Build GRCh38
rs375056989 Chr.11: 59192087-59192087 on Build GRCh38
rs776824955 Chr.11: 59192127-59192127 on Build GRCh38
rs199580019 Chr.11: 59192279-59192279 on Build GRCh38
rs112958827 Chr.11: 59192265-59192265 on Build GRCh38
rs186374111 Chr.11: 59192160-59192160 on Build GRCh38

View Full Product Details