Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25482707-25482964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTNB
Entrez Gene ID
1838
Gene Name
dystrobrevin beta
Gene Aliases
-
UniGene
Hs.307720
Gene Chromosome Location
Chr.2: 25377220-25673647 on Build GRCh38
Transcripts
NM_001256303.1 NM_001256304.1 NM_001256308.2 NM_001320932.1 NM_001320933.1 NM_001320934.1 NM_001320935.1 NM_001320936.1 NM_021907.4 NM_033147.3 NM_033148.3 NM_183360.2 NM_183361.2 XM_005264182.1 XM_011532671.1 XM_011532672.2 XM_011532673.1 XM_011532674.1 XM_011532675.1 XM_011532676.1 XM_011532677.2 XM_011532679.1 XM_011532680.2 XM_011532681.2 XM_017003529.1 XM_017003530.1 XM_017003531.1 XM_017003532.1 XM_017003533.1 XM_017003534.1 XM_017003535.1 XM_017003536.1 XM_017003537.1 XM_017003538.1 XM_017003539.1 XM_017003540.1 XM_017003541.1 XM_017003542.1 XM_017003543.1 XM_017003544.1 XM_017003545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6157728 c.1077G>C p.K359N Chr.2: 25482798-25482798 on Build GRCh38
4811825 c.1026G>A p.M342I Chr.2: 25482849-25482849 on Build GRCh38
4816806 c.1017G>A p.L339L Chr.2: 25482858-25482858 on Build GRCh38
6157729 c.1077G>C p.K359N Chr.2: 25482798-25482798 on Build GRCh38
5735971 c.1016T>C p.L339P Chr.2: 25482859-25482859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760188526 Chr.2: 25482792-25482792 on Build GRCh38
rs755758251 Chr.2: 25482782-25482782 on Build GRCh38
rs774689228 Chr.2: 25482767-25482767 on Build GRCh38
rs761145944 Chr.2: 25482898-25482898 on Build GRCh38
rs947666226 Chr.2: 25482936-25482936 on Build GRCh38
rs190445677 Chr.2: 25482865-25482865 on Build GRCh38
rs749552514 Chr.2: 25482863-25482863 on Build GRCh38
rs768070454 Chr.2: 25482888-25482888 on Build GRCh38
rs561030486 Chr.2: 25482743-25482743 on Build GRCh38
rs763873937 Chr.2: 25482792-25482792 on Build GRCh38
rs749762246 Chr.2: 25482760-25482760 on Build GRCh38
rs766494883 Chr.2: 25482762-25482762 on Build GRCh38
rs773235367 Chr.2: 25482752-25482752 on Build GRCh38
rs746813132 Chr.2: 25482847-25482847 on Build GRCh38
rs981185581 Chr.2: 25482827-25482827 on Build GRCh38
rs564400511 Chr.2: 25482736-25482736 on Build GRCh38
rs182089084 Chr.2: 25482866-25482866 on Build GRCh38
rs921553994 Chr.2: 25482915-25482915 on Build GRCh38
rs1050318054 Chr.2: 25482942-25482942 on Build GRCh38
rs984541765 Chr.2: 25482791-25482791 on Build GRCh38
rs767755718 Chr.2: 25482775-25482775 on Build GRCh38
rs145278466 Chr.2: 25482768-25482768 on Build GRCh38
rs368821580 Chr.2: 25482824-25482824 on Build GRCh38
rs759898782 Chr.2: 25482868-25482868 on Build GRCh38
rs757041263 Chr.2: 25482917-25482917 on Build GRCh38
rs779658231 Chr.2: 25482839-25482839 on Build GRCh38
rs201023279 Chr.2: 25482861-25482861 on Build GRCh38
rs775674865 Chr.2: 25482894-25482894 on Build GRCh38
rs764605554 Chr.2: 25482904-25482904 on Build GRCh38
rs903349779 Chr.2: 25482934-25482934 on Build GRCh38
rs752725257 Chr.2: 25482757-25482757 on Build GRCh38
rs756810355 Chr.2: 25482793-25482793 on Build GRCh38
rs769952609 Chr.2: 25482851-25482851 on Build GRCh38
rs908881614 Chr.2: 25482857-25482857 on Build GRCh38
rs116438487 Chr.2: 25482919-25482919 on Build GRCh38
rs565636505 Chr.2: 25482927-25482927 on Build GRCh38
rs758419981 Chr.2: 25482830-25482830 on Build GRCh38
rs778529061 Chr.2: 25482807-25482807 on Build GRCh38
rs368074459 Chr.2: 25482933-25482933 on Build GRCh38
rs538296431 Chr.2: 25482737-25482758 on Build GRCh38
rs777275806 Chr.2: 25482893-25482893 on Build GRCh38
rs1040359963 Chr.2: 25482889-25482889 on Build GRCh38
rs753115666 Chr.2: 25482779-25482779 on Build GRCh38

View Full Product Details