Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194425145-194425414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A3
Entrez Gene ID
79572
Gene Name
ATPase 13A3
Gene Aliases
AFURS1
UniGene
Hs.529609
Gene Chromosome Location
Chr.3: 194402674-194487006 on Build GRCh38
Transcripts
NM_024524.3 XM_005269357.3 XM_011513120.2 XM_011513121.1 XM_011513122.1 XM_011513123.2 XM_011513124.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3992948 c.3263C>T p.A1088V Chr.3: 194425392-194425392 on Build GRCh38
1168992 c.3271_3272insCCT p.A1090_F1091insS Chr.3: 194425383-194425384 on Build GRCh38
5440679 c.3275C>G p.S1092* Chr.3: 194425380-194425380 on Build GRCh38
7095198 c.3281G>T p.G1094V Chr.3: 194425374-194425374 on Build GRCh38
1159565 c.3270_3271insCCT p.A1090_F1091insP Chr.3: 194425384-194425385 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761132337 Chr.3: 194425243-194425243 on Build GRCh38
rs552422609 Chr.3: 194425246-194425246 on Build GRCh38
rs777206586 Chr.3: 194425311-194425311 on Build GRCh38
rs768328046 Chr.3: 194425221-194425221 on Build GRCh38
rs564690259 Chr.3: 194425268-194425268 on Build GRCh38
rs185763853 Chr.3: 194425232-194425232 on Build GRCh38
rs560135797 Chr.3: 194425206-194425206 on Build GRCh38
rs968471588 Chr.3: 194425332-194425332 on Build GRCh38
rs773825874 Chr.3: 194425237-194425237 on Build GRCh38
rs1011452306 Chr.3: 194425272-194425272 on Build GRCh38
rs117299962 Chr.3: 194425292-194425292 on Build GRCh38
rs1022717610 Chr.3: 194425326-194425326 on Build GRCh38
rs767334710 Chr.3: 194425298-194425298 on Build GRCh38
rs541785715 Chr.3: 194425172-194425172 on Build GRCh38
rs755825066 Chr.3: 194425329-194425329 on Build GRCh38
rs895708860 Chr.3: 194425290-194425290 on Build GRCh38
rs752577919 Chr.3: 194425300-194425300 on Build GRCh38
rs373987252 Chr.3: 194425347-194425347 on Build GRCh38
rs750613950 Chr.3: 194425367-194425367 on Build GRCh38
rs1021173682 Chr.3: 194425307-194425307 on Build GRCh38

View Full Product Details