Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56083706-56083971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERBB3
Entrez Gene ID
2065
Gene Name
erb-b2 receptor tyrosine kinase 3
Gene Aliases
ErbB-3, HER3, LCCS2, MDA-BF-1, c-erbB-3, c-erbB3, erbB3-S, p180-ErbB3, p45-sErbB3, p85-sErbB3
UniGene
Hs.118681
Gene Chromosome Location
Chr.12: 56080025-56103507 on Build GRCh38
Transcripts
NM_001005915.1 NM_001982.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6306065 c.167G>T p.C56F Chr.12: 56083835-56083835 on Build GRCh38
118305 c.147G>T p.L49L Chr.12: 56083815-56083815 on Build GRCh38
1606365 c.177G>T p.V59V Chr.12: 56083845-56083845 on Build GRCh38
6655801 c.199C>T p.L67F Chr.12: 56083867-56083867 on Build GRCh38
5611281 c.204_205GG>AA p.G69R Chr.12: 56083872-56083873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903404743 Chr.12: 56083737-56083737 on Build GRCh38
rs764891356 Chr.12: 56083812-56083812 on Build GRCh38
rs749691155 Chr.12: 56083849-56083849 on Build GRCh38
rs765693205 Chr.12: 56083884-56083884 on Build GRCh38
rs780212356 Chr.12: 56083843-56083843 on Build GRCh38
rs56017157 Chr.12: 56083757-56083757 on Build GRCh38
rs774536437 Chr.12: 56083786-56083786 on Build GRCh38
rs752412861 Chr.12: 56083909-56083909 on Build GRCh38
rs781661153 Chr.12: 56083831-56083831 on Build GRCh38
rs780636036 Chr.12: 56083949-56083949 on Build GRCh38
rs750947241 Chr.12: 56083840-56083840 on Build GRCh38
rs766147426 Chr.12: 56083795-56083795 on Build GRCh38
rs770120793 Chr.12: 56083765-56083765 on Build GRCh38
rs757449230 Chr.12: 56083917-56083917 on Build GRCh38
rs142735651 Chr.12: 56083871-56083871 on Build GRCh38
rs773189933 Chr.12: 56083878-56083878 on Build GRCh38
rs143770796 Chr.12: 56083885-56083885 on Build GRCh38
rs752180033 Chr.12: 56083826-56083826 on Build GRCh38
rs747687493 Chr.12: 56083752-56083752 on Build GRCh38
rs756535556 Chr.12: 56083931-56083931 on Build GRCh38
rs763532328 Chr.12: 56083913-56083913 on Build GRCh38
rs569378810 Chr.12: 56083731-56083731 on Build GRCh38
rs760399408 Chr.12: 56083872-56083872 on Build GRCh38
rs751083456 Chr.12: 56083915-56083915 on Build GRCh38
rs762010415 Chr.12: 56083788-56083788 on Build GRCh38
rs759044259 Chr.12: 56083811-56083811 on Build GRCh38
rs775764667 Chr.12: 56083767-56083767 on Build GRCh38
rs552030360 Chr.12: 56083851-56083851 on Build GRCh38
rs961946828 Chr.12: 56083907-56083907 on Build GRCh38
rs753681468 Chr.12: 56083806-56083806 on Build GRCh38
rs992281120 Chr.12: 56083827-56083827 on Build GRCh38
rs746211618 Chr.12: 56083758-56083758 on Build GRCh38
rs757941835 Chr.12: 56083828-56083828 on Build GRCh38
rs748210428 Chr.12: 56083869-56083869 on Build GRCh38
rs760515344 Chr.12: 56083792-56083792 on Build GRCh38
rs757712749 Chr.12: 56083753-56083753 on Build GRCh38
rs749981785 Chr.12: 56083865-56083880 on Build GRCh38
rs749408634 Chr.12: 56083785-56083785 on Build GRCh38
rs764836319 Chr.12: 56083904-56083904 on Build GRCh38
rs199806001 Chr.12: 56083899-56083899 on Build GRCh38
rs750029748 Chr.12: 56083783-56083783 on Build GRCh38
rs374953448 Chr.12: 56083789-56083789 on Build GRCh38
rs779330902 Chr.12: 56083862-56083862 on Build GRCh38
rs570614098 Chr.12: 56083883-56083883 on Build GRCh38
rs756656037 Chr.12: 56083842-56083842 on Build GRCh38
rs764925612 Chr.12: 56083814-56083815 on Build GRCh38
rs2271194 Chr.12: 56083910-56083910 on Build GRCh38

View Full Product Details