Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62566751-62567024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEF1G
Entrez Gene ID
1937
Gene Name
eukaryotic translation elongation factor 1 gamma
Gene Aliases
EF1G, GIG35
UniGene
Hs.144835
Gene Chromosome Location
Chr.11: 62559601-62573988 on Build GRCh38
Transcripts
NM_001404.4 NR_106805.1

Gene Symbol
MIR6747
Entrez Gene ID
102465447
Gene Name
microRNA 6747
Gene Aliases
hsa-mir-6747
UniGene
Gene Chromosome Location
Chr.11: 62567011-62567071 on Build GRCh38
Transcripts
NM_001404.4 NR_106805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4198834 c.917C>T p.A306V Chr.11: 62566896-62566896 on Build GRCh38
4198835 c.908A>G p.K303R Chr.11: 62566905-62566905 on Build GRCh38
6423877 c.943G>T p.E315* Chr.11: 62566870-62566870 on Build GRCh38
1250957 c.695G>A p.R232Q Chr.11: 62566968-62566968 on Build GRCh38
7194678 c.661T>C p.F221L Chr.11: 62567002-62567002 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747786772 Chr.11: 62566957-62566957 on Build GRCh38
rs746771975 Chr.11: 62566970-62566970 on Build GRCh38
rs781687410 Chr.11: 62566920-62566920 on Build GRCh38
rs1010231736 Chr.11: 62566943-62566943 on Build GRCh38
rs199610030 Chr.11: 62566892-62566892 on Build GRCh38
rs943526549 Chr.11: 62566789-62566789 on Build GRCh38
rs371597439 Chr.11: 62566988-62566988 on Build GRCh38
rs771965894 Chr.11: 62566823-62566823 on Build GRCh38
rs374645303 Chr.11: 62566958-62566958 on Build GRCh38
rs759624665 Chr.11: 62566872-62566872 on Build GRCh38
rs371308314 Chr.11: 62566921-62566921 on Build GRCh38
rs1047756579 Chr.11: 62566867-62566867 on Build GRCh38
rs761595268 Chr.11: 62566821-62566821 on Build GRCh38
rs11545771 Chr.11: 62566990-62566990 on Build GRCh38
rs772756652 Chr.11: 62566969-62566969 on Build GRCh38
rs758677352 Chr.11: 62566893-62566893 on Build GRCh38
rs778611401 Chr.11: 62566951-62566951 on Build GRCh38
rs1038272444 Chr.11: 62566931-62566931 on Build GRCh38
rs923344557 Chr.11: 62566822-62566822 on Build GRCh38
rs933785027 Chr.11: 62566807-62566807 on Build GRCh38
rs11549631 Chr.11: 62566992-62566992 on Build GRCh38
rs748772853 Chr.11: 62566809-62566809 on Build GRCh38
rs367830508 Chr.11: 62566901-62566901 on Build GRCh38
rs760522344 Chr.11: 62566854-62566854 on Build GRCh38
rs11545770 Chr.11: 62566997-62566997 on Build GRCh38
rs368054471 Chr.11: 62566974-62566974 on Build GRCh38
rs770666889 Chr.11: 62566973-62566973 on Build GRCh38
rs372879019 Chr.11: 62566857-62566857 on Build GRCh38
rs371535693 Chr.11: 62566797-62566797 on Build GRCh38
rs768249627 Chr.11: 62566932-62566932 on Build GRCh38
rs757574790 Chr.11: 62566902-62566902 on Build GRCh38
rs766285337 Chr.11: 62566856-62566856 on Build GRCh38
rs773114543 Chr.11: 62566830-62566830 on Build GRCh38
rs774106674 Chr.11: 62566819-62566819 on Build GRCh38
rs775985944 Chr.11: 62566779-62566780 on Build GRCh38
rs768303087 Chr.11: 62566816-62566816 on Build GRCh38
rs865829861 Chr.11: 62566998-62566998 on Build GRCh38

View Full Product Details