Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33578240-33578501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DONSON
Entrez Gene ID
29980
Gene Name
downstream neighbor of SON
Gene Aliases
B17, C21orf60
UniGene
Hs.436341
Gene Chromosome Location
Chr.21: 33577551-33588708 on Build GRCh38
Transcripts
NM_017613.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5392839 c.1564-1G>A p.? Chr.21: 33578445-33578445 on Build GRCh38
6325008 c.1691G>A p.W564* Chr.21: 33578317-33578317 on Build GRCh38
1725180 c.1675G>C p.D559H Chr.21: 33578333-33578333 on Build GRCh38
1413902 c.1700G>A p.*567* Chr.21: 33578308-33578308 on Build GRCh38
1659576 c.1691G>C p.W564S Chr.21: 33578317-33578317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772168759 Chr.21: 33578378-33578378 on Build GRCh38
rs746497389 Chr.21: 33578445-33578446 on Build GRCh38
rs774616573 Chr.21: 33578374-33578374 on Build GRCh38
rs941110182 Chr.21: 33578340-33578340 on Build GRCh38
rs775449454 Chr.21: 33578383-33578383 on Build GRCh38
rs778412315 Chr.21: 33578450-33578450 on Build GRCh38
rs776282583 Chr.21: 33578399-33578399 on Build GRCh38
rs763879389 Chr.21: 33578398-33578398 on Build GRCh38
rs924451011 Chr.21: 33578439-33578439 on Build GRCh38
rs940080293 Chr.21: 33578329-33578329 on Build GRCh38
rs778013273 Chr.21: 33578352-33578352 on Build GRCh38
rs751648673 Chr.21: 33578387-33578387 on Build GRCh38
rs771138584 Chr.21: 33578373-33578373 on Build GRCh38
rs758034238 Chr.21: 33578349-33578349 on Build GRCh38
rs749937330 Chr.21: 33578338-33578338 on Build GRCh38
rs756584370 Chr.21: 33578434-33578434 on Build GRCh38
rs147521584 Chr.21: 33578393-33578393 on Build GRCh38
rs753472885 Chr.21: 33578311-33578311 on Build GRCh38
rs529567515 Chr.21: 33578404-33578404 on Build GRCh38
rs1012950140 Chr.21: 33578342-33578342 on Build GRCh38
rs745916219 Chr.21: 33578375-33578375 on Build GRCh38
rs765045326 Chr.21: 33578337-33578337 on Build GRCh38
rs752614295 Chr.21: 33578297-33578297 on Build GRCh38
rs141237312 Chr.21: 33578422-33578422 on Build GRCh38
rs777467660 Chr.21: 33578278-33578279 on Build GRCh38
rs749446485 Chr.21: 33578359-33578359 on Build GRCh38
rs957480991 Chr.21: 33578440-33578440 on Build GRCh38
rs1003541624 Chr.21: 33578391-33578391 on Build GRCh38
rs749722029 Chr.21: 33578420-33578420 on Build GRCh38
rs748637250 Chr.21: 33578350-33578350 on Build GRCh38
rs768393448 Chr.21: 33578414-33578415 on Build GRCh38
rs752704331 Chr.21: 33578267-33578268 on Build GRCh38
rs113553347 Chr.21: 33578351-33578351 on Build GRCh38
rs528633155 Chr.21: 33578275-33578275 on Build GRCh38
rs757031355 Chr.21: 33578454-33578455 on Build GRCh38
rs764959235 Chr.21: 33578405-33578405 on Build GRCh38
rs951519695 Chr.21: 33578292-33578292 on Build GRCh38
rs756970023 Chr.21: 33578332-33578332 on Build GRCh38
rs758229003 Chr.21: 33578277-33578278 on Build GRCh38
rs116461921 Chr.21: 33578303-33578303 on Build GRCh38

View Full Product Details