Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 175879815-175880086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPLX2
Entrez Gene ID
10814
Gene Name
complexin 2
Gene Aliases
921-L, CPX-2, CPX2, Hfb1
UniGene
Hs.193235
Gene Chromosome Location
Chr.5: 175796539-175884021 on Build GRCh38
Transcripts
NM_001008220.1 NM_006650.3 XM_005265798.1 XM_005265799.1 XM_011534419.1 XM_017008964.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6012149 c.241G>A p.E81K Chr.5: 175879881-175879881 on Build GRCh38
4337880 c.322_324delGAG p.E114delE Chr.5: 175879962-175879964 on Build GRCh38
1310983 c.274G>C p.G92R Chr.5: 175879914-175879914 on Build GRCh38
5868182 c.304C>T p.P102S Chr.5: 175879944-175879944 on Build GRCh38
1065969 c.305C>A p.P102H Chr.5: 175879945-175879945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745515602 Chr.5: 175879840-175879840 on Build GRCh38
rs547842894 Chr.5: 175879961-175879961 on Build GRCh38
rs746469867 Chr.5: 175879878-175879878 on Build GRCh38
rs964944553 Chr.5: 175880004-175880004 on Build GRCh38
rs771751735 Chr.5: 175879869-175879869 on Build GRCh38
rs746666225 Chr.5: 175879988-175879988 on Build GRCh38
rs755639183 Chr.5: 175879913-175879913 on Build GRCh38
rs759810852 Chr.5: 175879921-175879921 on Build GRCh38
rs772923966 Chr.5: 175879911-175879911 on Build GRCh38
rs956440635 Chr.5: 175880022-175880022 on Build GRCh38
rs760771461 Chr.5: 175879948-175879948 on Build GRCh38
rs565621803 Chr.5: 175880021-175880021 on Build GRCh38
rs753000048 Chr.5: 175879931-175879931 on Build GRCh38
rs745994665 Chr.5: 175879961-175879962 on Build GRCh38
rs770855795 Chr.5: 175880052-175880052 on Build GRCh38
rs770953273 Chr.5: 175879864-175879866 on Build GRCh38
rs777361096 Chr.5: 175880016-175880016 on Build GRCh38
rs267600558 Chr.5: 175879945-175879945 on Build GRCh38
rs946484429 Chr.5: 175879957-175879957 on Build GRCh38
rs776563428 Chr.5: 175879895-175879895 on Build GRCh38
rs368317661 Chr.5: 175879999-175879999 on Build GRCh38
rs183694640 Chr.5: 175879964-175879964 on Build GRCh38
rs758781221 Chr.5: 175879968-175879968 on Build GRCh38
rs768201279 Chr.5: 175879990-175879990 on Build GRCh38
rs1047967443 Chr.5: 175879959-175879959 on Build GRCh38
rs560271013 Chr.5: 175880001-175880001 on Build GRCh38
rs761717452 Chr.5: 175879904-175879904 on Build GRCh38
rs750831279 Chr.5: 175879967-175879967 on Build GRCh38
rs544929121 Chr.5: 175879910-175879910 on Build GRCh38
rs757770941 Chr.5: 175879956-175879956 on Build GRCh38
rs774954265 Chr.5: 175879875-175879875 on Build GRCh38
rs779201569 Chr.5: 175879833-175879833 on Build GRCh38
rs150632571 Chr.5: 175879949-175879949 on Build GRCh38
rs776387890 Chr.5: 175879962-175879964 on Build GRCh38
rs1007680810 Chr.5: 175880058-175880058 on Build GRCh38
rs555416420 Chr.5: 175880057-175880057 on Build GRCh38
rs780637203 Chr.5: 175879997-175879997 on Build GRCh38
rs769672063 Chr.5: 175880015-175880015 on Build GRCh38
rs374428734 Chr.5: 175879972-175879972 on Build GRCh38
rs934850700 Chr.5: 175879880-175879880 on Build GRCh38
rs370394171 Chr.5: 175879929-175879929 on Build GRCh38
rs768079195 Chr.5: 175879886-175879886 on Build GRCh38

View Full Product Details