Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48771751-48772008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY6
Entrez Gene ID
112
Gene Name
adenylate cyclase 6
Gene Aliases
AC6, LCCS8
UniGene
Hs.525401
Gene Chromosome Location
Chr.12: 48766191-48789096 on Build GRCh38
Transcripts
NM_015270.4 XM_006719210.3 XM_017018743.1 NR_039850.2

Gene Symbol
MIR4701
Entrez Gene ID
100616262
Gene Name
microRNA 4701
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.12: 48771975-48772037 on Build GRCh38
Transcripts
NM_015270.4 XM_006719210.3 XM_017018743.1 NR_039850.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6340826 c.2994C>A p.N998K Chr.12: 48771767-48771767 on Build GRCh38
4659929 c.2896C>T p.R966C Chr.12: 48771865-48771865 on Build GRCh38
5495439 c.2820A>T p.L940L Chr.12: 48771941-48771941 on Build GRCh38
7088013 c.2900A>G p.N967S Chr.12: 48771861-48771861 on Build GRCh38
1676924 c.2869G>A p.A957T Chr.12: 48771892-48771892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368049910 Chr.12: 48771822-48771823 on Build GRCh38
rs754462882 Chr.12: 48771907-48771907 on Build GRCh38
rs906075876 Chr.12: 48771912-48771912 on Build GRCh38
rs764365272 Chr.12: 48771868-48771868 on Build GRCh38
rs143779554 Chr.12: 48771806-48771806 on Build GRCh38
rs563955257 Chr.12: 48771849-48771849 on Build GRCh38
rs746847957 Chr.12: 48771962-48771962 on Build GRCh38
rs755219383 Chr.12: 48771967-48771968 on Build GRCh38
rs778563944 Chr.12: 48771927-48771927 on Build GRCh38
rs143958339 Chr.12: 48771867-48771867 on Build GRCh38
rs1000937427 Chr.12: 48771860-48771860 on Build GRCh38
rs756654985 Chr.12: 48771889-48771889 on Build GRCh38
rs779684053 Chr.12: 48771791-48771791 on Build GRCh38
rs778485526 Chr.12: 48771839-48771839 on Build GRCh38
rs778196578 Chr.12: 48771869-48771869 on Build GRCh38
rs985391505 Chr.12: 48771861-48771861 on Build GRCh38
rs777523139 Chr.12: 48771944-48771944 on Build GRCh38
rs768346518 Chr.12: 48771819-48771819 on Build GRCh38
rs774582629 Chr.12: 48771979-48771979 on Build GRCh38
rs750863326 Chr.12: 48771884-48771884 on Build GRCh38
rs200640184 Chr.12: 48771968-48771968 on Build GRCh38
rs932653682 Chr.12: 48771818-48771818 on Build GRCh38
rs747790458 Chr.12: 48771932-48771932 on Build GRCh38
rs766252180 Chr.12: 48771851-48771852 on Build GRCh38
rs200070441 Chr.12: 48771840-48771840 on Build GRCh38
rs776609767 Chr.12: 48771964-48771964 on Build GRCh38
rs762784516 Chr.12: 48771793-48771794 on Build GRCh38
rs1023470388 Chr.12: 48771820-48771820 on Build GRCh38
rs753383547 Chr.12: 48771896-48771896 on Build GRCh38
rs755635840 Chr.12: 48771787-48771787 on Build GRCh38
rs748843958 Chr.12: 48771807-48771807 on Build GRCh38
rs770852283 Chr.12: 48771963-48771963 on Build GRCh38
rs751732732 Chr.12: 48771873-48771873 on Build GRCh38
rs771856644 Chr.12: 48771935-48771935 on Build GRCh38
rs760633041 Chr.12: 48771864-48771864 on Build GRCh38
rs747921948 Chr.12: 48771844-48771844 on Build GRCh38
rs897307392 Chr.12: 48771880-48771880 on Build GRCh38
rs147278695 Chr.12: 48771965-48771965 on Build GRCh38
rs368945637 Chr.12: 48771874-48771874 on Build GRCh38
rs773096680 Chr.12: 48771851-48771851 on Build GRCh38
rs138465901 Chr.12: 48771865-48771865 on Build GRCh38
rs540786987 Chr.12: 48771832-48771832 on Build GRCh38
rs879253864 Chr.12: 48771786-48771786 on Build GRCh38
rs538084003 Chr.12: 48771983-48771983 on Build GRCh38
rs563129186 Chr.12: 48771876-48771876 on Build GRCh38
rs371352626 Chr.12: 48771890-48771890 on Build GRCh38
rs751347638 Chr.12: 48771912-48771913 on Build GRCh38

View Full Product Details