Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27756582-27756843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
UniGene
Hs.445512
Gene Chromosome Location
Chr.8: 27733316-27772656 on Build GRCh38
Transcripts
NM_001304529.1 NM_001304530.1 NM_001304532.1 NM_018246.2 NR_130761.1 NR_130762.1 XM_011544570.2 XM_011544571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274411 c.157C>T p.R53* Chr.8: 27756730-27756730 on Build GRCh38
1098781 c.158G>A p.R53Q Chr.8: 27756729-27756729 on Build GRCh38
3925107 c.127G>A p.D43N Chr.8: 27756760-27756760 on Build GRCh38
1098782 c.120T>G p.F40L Chr.8: 27756767-27756767 on Build GRCh38
3647853 c.154C>T p.L52F Chr.8: 27756733-27756733 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779085297 Chr.8: 27756755-27756755 on Build GRCh38
rs2280884 Chr.8: 27756621-27756621 on Build GRCh38
rs776490576 Chr.8: 27756676-27756676 on Build GRCh38
rs372611484 Chr.8: 27756648-27756648 on Build GRCh38
rs754907988 Chr.8: 27756684-27756684 on Build GRCh38
rs748940794 Chr.8: 27756741-27756741 on Build GRCh38
rs1047749103 Chr.8: 27756639-27756639 on Build GRCh38
rs747557723 Chr.8: 27756730-27756730 on Build GRCh38
rs888838295 Chr.8: 27756615-27756615 on Build GRCh38
rs764169558 Chr.8: 27756675-27756675 on Build GRCh38
rs146456175 Chr.8: 27756682-27756682 on Build GRCh38
rs768891007 Chr.8: 27756749-27756750 on Build GRCh38
rs12543169 Chr.8: 27756620-27756620 on Build GRCh38
rs900670181 Chr.8: 27756796-27756796 on Build GRCh38
rs990865785 Chr.8: 27756616-27756616 on Build GRCh38
rs996976286 Chr.8: 27756813-27756813 on Build GRCh38
rs1038241682 Chr.8: 27756721-27756721 on Build GRCh38
rs774851097 Chr.8: 27756774-27756774 on Build GRCh38
rs777250706 Chr.8: 27756737-27756737 on Build GRCh38
rs746237749 Chr.8: 27756805-27756805 on Build GRCh38
rs367645106 Chr.8: 27756746-27756746 on Build GRCh38
rs370283861 Chr.8: 27756817-27756817 on Build GRCh38
rs770856547 Chr.8: 27756627-27756627 on Build GRCh38
rs370725443 Chr.8: 27756669-27756669 on Build GRCh38
rs202223241 Chr.8: 27756734-27756734 on Build GRCh38
rs886674368 Chr.8: 27756653-27756653 on Build GRCh38
rs781116567 Chr.8: 27756696-27756696 on Build GRCh38
rs751416490 Chr.8: 27756683-27756683 on Build GRCh38
rs200313388 Chr.8: 27756679-27756679 on Build GRCh38

View Full Product Details