Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33521366-33521633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
UniGene
Hs.656915
Gene Chromosome Location
Chr.1: 33513999-34165842 on Build GRCh38
Transcripts
NM_001281956.1 NM_052896.4 XM_017000185.1 XM_017000186.1 XM_017000187.1 XM_017000188.1 XM_017000189.1 XM_017000190.1 XM_017000194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1646181 c.10561G>T p.G3521C Chr.1: 33521499-33521499 on Build GRCh38
5603299 c.10514C>T p.S3505L Chr.1: 33521546-33521546 on Build GRCh38
681055 c.10117G>A p.G3373S Chr.1: 33521511-33521511 on Build GRCh38
6636406 c.10112A>G p.Y3371C Chr.1: 33521516-33521516 on Build GRCh38
1646180 c.10549G>A p.G3517S Chr.1: 33521511-33521511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746146936 Chr.1: 33521478-33521478 on Build GRCh38
rs2641949 Chr.1: 33521441-33521441 on Build GRCh38
rs1023956732 Chr.1: 33521579-33521579 on Build GRCh38
rs765381300 Chr.1: 33521613-33521613 on Build GRCh38
rs888257633 Chr.1: 33521505-33521505 on Build GRCh38
rs762678456 Chr.1: 33521422-33521422 on Build GRCh38
rs750437715 Chr.1: 33521546-33521546 on Build GRCh38
rs757320710 Chr.1: 33521447-33521447 on Build GRCh38
rs372572008 Chr.1: 33521573-33521573 on Build GRCh38
rs747236467 Chr.1: 33521571-33521571 on Build GRCh38
rs936450610 Chr.1: 33521393-33521393 on Build GRCh38
rs201777176 Chr.1: 33521585-33521585 on Build GRCh38
rs555477246 Chr.1: 33521499-33521499 on Build GRCh38
rs766087881 Chr.1: 33521506-33521506 on Build GRCh38
rs769231175 Chr.1: 33521489-33521489 on Build GRCh38
rs762696646 Chr.1: 33521503-33521503 on Build GRCh38
rs752644843 Chr.1: 33521537-33521537 on Build GRCh38
rs767311659 Chr.1: 33521534-33521534 on Build GRCh38
rs372754000 Chr.1: 33521429-33521429 on Build GRCh38
rs368499050 Chr.1: 33521428-33521428 on Build GRCh38
rs375873007 Chr.1: 33521446-33521446 on Build GRCh38
rs374814024 Chr.1: 33521575-33521575 on Build GRCh38
rs780203488 Chr.1: 33521569-33521569 on Build GRCh38
rs780403208 Chr.1: 33521485-33521485 on Build GRCh38
rs1049162205 Chr.1: 33521508-33521508 on Build GRCh38
rs960922700 Chr.1: 33521572-33521572 on Build GRCh38
rs747298393 Chr.1: 33521486-33521486 on Build GRCh38
rs751420152 Chr.1: 33521532-33521532 on Build GRCh38
rs1773028 Chr.1: 33521458-33521458 on Build GRCh38
rs778961286 Chr.1: 33521449-33521449 on Build GRCh38
rs2794595 Chr.1: 33521524-33521524 on Build GRCh38
rs756066303 Chr.1: 33521545-33521545 on Build GRCh38
rs1690547 Chr.1: 33521578-33521578 on Build GRCh38
rs777058143 Chr.1: 33521490-33521490 on Build GRCh38
rs987828031 Chr.1: 33521399-33521399 on Build GRCh38
rs772787986 Chr.1: 33521501-33521501 on Build GRCh38
rs913585673 Chr.1: 33521409-33521409 on Build GRCh38
rs763606278 Chr.1: 33521567-33521568 on Build GRCh38
rs368867925 Chr.1: 33521498-33521498 on Build GRCh38
rs1690548 Chr.1: 33521603-33521603 on Build GRCh38
rs776542804 Chr.1: 33521535-33521535 on Build GRCh38
rs759298187 Chr.1: 33521533-33521533 on Build GRCh38
rs762354229 Chr.1: 33521492-33521492 on Build GRCh38
rs202198885 Chr.1: 33521427-33521427 on Build GRCh38
rs762003017 Chr.1: 33521568-33521568 on Build GRCh38
rs780942043 Chr.1: 33521442-33521442 on Build GRCh38
rs1039457650 Chr.1: 33521448-33521448 on Build GRCh38
rs772225864 Chr.1: 33521479-33521479 on Build GRCh38

View Full Product Details