Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100550347-100550572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG2
Entrez Gene ID
3268
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
HRBL, RABR
UniGene
Hs.521083
Gene Chromosome Location
Chr.7: 100539211-100568220 on Build GRCh38
Transcripts
NM_006076.4 XM_005250306.2 XM_017012112.1 XM_017012113.1 XM_017012114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
172591 c.327G>T p.K109N Chr.7: 100550407-100550407 on Build GRCh38
1082864 c.414A>T p.K138N Chr.7: 100550494-100550494 on Build GRCh38
5946454 c.347A>T p.D116V Chr.7: 100550427-100550427 on Build GRCh38
4517573 c.374_375CC>TT p.S125F Chr.7: 100550454-100550455 on Build GRCh38
6596231 c.377G>A p.R126K Chr.7: 100550457-100550457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770694129 Chr.7: 100550543-100550546 on Build GRCh38
rs370191289 Chr.7: 100550517-100550517 on Build GRCh38
rs748556561 Chr.7: 100550475-100550475 on Build GRCh38
rs770552533 Chr.7: 100550507-100550507 on Build GRCh38
rs771970852 Chr.7: 100550519-100550519 on Build GRCh38
rs775536622 Chr.7: 100550529-100550529 on Build GRCh38
rs762503806 Chr.7: 100550385-100550385 on Build GRCh38
rs781652659 Chr.7: 100550462-100550462 on Build GRCh38
rs750650620 Chr.7: 100550433-100550433 on Build GRCh38
rs767431926 Chr.7: 100550403-100550403 on Build GRCh38
rs753644967 Chr.7: 100550543-100550543 on Build GRCh38
rs748973775 Chr.7: 100550494-100550494 on Build GRCh38
rs758596353 Chr.7: 100550436-100550436 on Build GRCh38
rs375257050 Chr.7: 100550483-100550483 on Build GRCh38
rs948775820 Chr.7: 100550447-100550447 on Build GRCh38
rs893877093 Chr.7: 100550369-100550369 on Build GRCh38
rs755449202 Chr.7: 100550446-100550446 on Build GRCh38
rs367548306 Chr.7: 100550389-100550389 on Build GRCh38
rs372318437 Chr.7: 100550537-100550537 on Build GRCh38
rs777713041 Chr.7: 100550383-100550384 on Build GRCh38
rs556715227 Chr.7: 100550414-100550414 on Build GRCh38
rs773875205 Chr.7: 100550393-100550393 on Build GRCh38
rs760595528 Chr.7: 100550533-100550533 on Build GRCh38
rs772823759 Chr.7: 100550370-100550370 on Build GRCh38
rs765595747 Chr.7: 100550432-100550432 on Build GRCh38
rs773749861 Chr.7: 100550511-100550511 on Build GRCh38
rs966666193 Chr.7: 100550489-100550489 on Build GRCh38
rs747041982 Chr.7: 100550439-100550439 on Build GRCh38
rs375827427 Chr.7: 100550437-100550437 on Build GRCh38
rs146728315 Chr.7: 100550402-100550402 on Build GRCh38
rs746865620 Chr.7: 100550510-100550511 on Build GRCh38
rs755967504 Chr.7: 100550430-100550430 on Build GRCh38
rs144744845 Chr.7: 100550400-100550400 on Build GRCh38

View Full Product Details