Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43155672-43155927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSGALNACT2
Entrez Gene ID
55454
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 2
Gene Aliases
CHGN2, ChGn-2, GALNACT-2, GALNACT2, PRO0082, beta4GalNAcT
UniGene
Hs.657569
Gene Chromosome Location
Chr.10: 43138445-43185308 on Build GRCh38
Transcripts
NM_001319654.1 NM_001319656.1 NM_018590.4 NR_135077.1 XM_011539938.1 XM_017016388.1 XM_017016389.1 XM_017016390.1 XM_017016391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
171900 c.654C>T p.F218F Chr.10: 43155803-43155803 on Build GRCh38
6636163 c.589G>C p.D197H Chr.10: 43155738-43155738 on Build GRCh38
6993239 c.610G>A p.E204K Chr.10: 43155759-43155759 on Build GRCh38
3665732 c.636A>G p.I212M Chr.10: 43155785-43155785 on Build GRCh38
5753005 c.599A>G p.Q200R Chr.10: 43155748-43155748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772123115 Chr.10: 43155826-43155826 on Build GRCh38
rs776867198 Chr.10: 43155845-43155845 on Build GRCh38
rs370758517 Chr.10: 43155865-43155865 on Build GRCh38
rs759419731 Chr.10: 43155722-43155722 on Build GRCh38
rs769418915 Chr.10: 43155701-43155701 on Build GRCh38
rs377323289 Chr.10: 43155855-43155855 on Build GRCh38
rs138631109 Chr.10: 43155700-43155700 on Build GRCh38
rs961578378 Chr.10: 43155758-43155758 on Build GRCh38
rs1026946231 Chr.10: 43155739-43155739 on Build GRCh38
rs757323470 Chr.10: 43155754-43155754 on Build GRCh38
rs181850949 Chr.10: 43155863-43155863 on Build GRCh38
rs144196824 Chr.10: 43155804-43155804 on Build GRCh38
rs752537770 Chr.10: 43155729-43155729 on Build GRCh38
rs754850765 Chr.10: 43155748-43155750 on Build GRCh38
rs764448693 Chr.10: 43155841-43155841 on Build GRCh38
rs548924168 Chr.10: 43155816-43155816 on Build GRCh38
rs141131798 Chr.10: 43155764-43155764 on Build GRCh38
rs750872809 Chr.10: 43155759-43155759 on Build GRCh38
rs759937265 Chr.10: 43155743-43155743 on Build GRCh38
rs758899799 Chr.10: 43155762-43155762 on Build GRCh38
rs868121389 Chr.10: 43155707-43155707 on Build GRCh38
rs199917944 Chr.10: 43155766-43155766 on Build GRCh38
rs767355897 Chr.10: 43155725-43155725 on Build GRCh38
rs764359099 Chr.10: 43155740-43155740 on Build GRCh38
rs761037262 Chr.10: 43155731-43155731 on Build GRCh38
rs764257096 Chr.10: 43155843-43155843 on Build GRCh38
rs199874536 Chr.10: 43155756-43155756 on Build GRCh38
rs915767770 Chr.10: 43155874-43155874 on Build GRCh38
rs879825703 Chr.10: 43155720-43155722 on Build GRCh38
rs778854932 Chr.10: 43155781-43155782 on Build GRCh38
rs777440588 Chr.10: 43155780-43155780 on Build GRCh38
rs11238456 Chr.10: 43155792-43155792 on Build GRCh38
rs770803776 Chr.10: 43155713-43155713 on Build GRCh38
rs753158194 Chr.10: 43155760-43155760 on Build GRCh38
rs984496209 Chr.10: 43155874-43155878 on Build GRCh38
rs1008772601 Chr.10: 43155738-43155738 on Build GRCh38
rs111984309 Chr.10: 43155885-43155885 on Build GRCh38
rs774294643 Chr.10: 43155720-43155720 on Build GRCh38
rs41298785 Chr.10: 43155895-43155895 on Build GRCh38
rs747352234 Chr.10: 43155768-43155768 on Build GRCh38
rs774161136 Chr.10: 43155815-43155815 on Build GRCh38
rs755803586 Chr.10: 43155773-43155773 on Build GRCh38
rs12571369 Chr.10: 43155782-43155782 on Build GRCh38
rs760121385 Chr.10: 43155706-43155706 on Build GRCh38
rs190429465 Chr.10: 43155838-43155838 on Build GRCh38
rs772646440 Chr.10: 43155704-43155704 on Build GRCh38
rs765422331 Chr.10: 43155862-43155862 on Build GRCh38

View Full Product Details