Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230660647-230660872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG2
Entrez Gene ID
22796
Gene Name
component of oligomeric golgi complex 2
Gene Aliases
LDLC
UniGene
Hs.211800
Gene Chromosome Location
Chr.1: 230642456-230693985 on Build GRCh38
Transcripts
NM_001145036.1 NM_007357.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
905365 c.243G>T p.M81I Chr.1: 230660766-230660766 on Build GRCh38
679134 c.245A>G p.D82G Chr.1: 230660768-230660768 on Build GRCh38
6632197 c.287G>A p.R96Q Chr.1: 230660810-230660810 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766625647 Chr.1: 230660752-230660753 on Build GRCh38
rs760604077 Chr.1: 230660741-230660741 on Build GRCh38
rs759567085 Chr.1: 230660714-230660714 on Build GRCh38
rs922690609 Chr.1: 230660690-230660690 on Build GRCh38
rs750707744 Chr.1: 230660783-230660783 on Build GRCh38
rs538382749 Chr.1: 230660784-230660784 on Build GRCh38
rs751763497 Chr.1: 230660796-230660796 on Build GRCh38
rs754168963 Chr.1: 230660765-230660765 on Build GRCh38
rs759212065 Chr.1: 230660833-230660833 on Build GRCh38
rs762582620 Chr.1: 230660707-230660707 on Build GRCh38
rs769246383 Chr.1: 230660791-230660791 on Build GRCh38
rs757543910 Chr.1: 230660774-230660774 on Build GRCh38
rs774153345 Chr.1: 230660818-230660818 on Build GRCh38
rs371730085 Chr.1: 230660834-230660834 on Build GRCh38
rs777262306 Chr.1: 230660795-230660795 on Build GRCh38
rs138501518 Chr.1: 230660790-230660790 on Build GRCh38
rs779456491 Chr.1: 230660775-230660775 on Build GRCh38
rs16852160 Chr.1: 230660787-230660787 on Build GRCh38
rs767308081 Chr.1: 230660719-230660719 on Build GRCh38
rs977279274 Chr.1: 230660681-230660681 on Build GRCh38
rs925934986 Chr.1: 230660793-230660793 on Build GRCh38
rs764421853 Chr.1: 230660759-230660759 on Build GRCh38
rs774102245 Chr.1: 230660708-230660708 on Build GRCh38
rs375004999 Chr.1: 230660835-230660835 on Build GRCh38
rs770549699 Chr.1: 230660805-230660805 on Build GRCh38
rs748979981 Chr.1: 230660802-230660802 on Build GRCh38
rs367979438 Chr.1: 230660726-230660726 on Build GRCh38

View Full Product Details