Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49794529-49794802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR4
Entrez Gene ID
389118
Gene Name
cadherin related family member 4
Gene Aliases
CDH29, PRO34300
UniGene
Hs.363312
Gene Chromosome Location
Chr.3: 49790732-49802741 on Build GRCh38
Transcripts
NM_001007540.3 XM_011533700.2 XM_011533701.2 XM_017006366.1 XM_017006367.1 XM_017006368.1 XM_017006369.1 XM_017006370.1 XM_017006371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6351794 c.1254C>G p.L418L Chr.3: 49794633-49794633 on Build GRCh38
6410216 c.1246C>A p.L416M Chr.3: 49794641-49794641 on Build GRCh38
6396815 c.1210A>G p.T404A Chr.3: 49794677-49794677 on Build GRCh38
5399612 c.1242C>T p.S414S Chr.3: 49794645-49794645 on Build GRCh38
5524728 c.1272G>A p.Q424Q Chr.3: 49794615-49794615 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778347547 Chr.3: 49794740-49794740 on Build GRCh38
rs778368316 Chr.3: 49794650-49794650 on Build GRCh38
rs911735116 Chr.3: 49794560-49794560 on Build GRCh38
rs772835403 Chr.3: 49794697-49794697 on Build GRCh38
rs373161672 Chr.3: 49794591-49794591 on Build GRCh38
rs1056370729 Chr.3: 49794630-49794630 on Build GRCh38
rs944397819 Chr.3: 49794563-49794563 on Build GRCh38
rs535014840 Chr.3: 49794771-49794771 on Build GRCh38
rs749895196 Chr.3: 49794652-49794652 on Build GRCh38
rs377295673 Chr.3: 49794687-49794687 on Build GRCh38
rs570154679 Chr.3: 49794592-49794592 on Build GRCh38
rs757873468 Chr.3: 49794674-49794674 on Build GRCh38
rs1047168034 Chr.3: 49794582-49794582 on Build GRCh38
rs570710863 Chr.3: 49794762-49794762 on Build GRCh38
rs762442754 Chr.3: 49794742-49794742 on Build GRCh38
rs768148707 Chr.3: 49794631-49794632 on Build GRCh38
rs756485912 Chr.3: 49794738-49794738 on Build GRCh38
rs752609821 Chr.3: 49794584-49794584 on Build GRCh38
rs529086299 Chr.3: 49794548-49794548 on Build GRCh38
rs550339619 Chr.3: 49794555-49794555 on Build GRCh38
rs919928959 Chr.3: 49794554-49794554 on Build GRCh38
rs868478851 Chr.3: 49794746-49794746 on Build GRCh38
rs117794954 Chr.3: 49794633-49794633 on Build GRCh38
rs757929086 Chr.3: 49794702-49794702 on Build GRCh38
rs759369620 Chr.3: 49794569-49794569 on Build GRCh38
rs1045088612 Chr.3: 49794667-49794667 on Build GRCh38
rs767440509 Chr.3: 49794576-49794576 on Build GRCh38
rs552551358 Chr.3: 49794644-49794644 on Build GRCh38
rs1012184412 Chr.3: 49794663-49794663 on Build GRCh38
rs888734651 Chr.3: 49794713-49794713 on Build GRCh38
rs1048621901 Chr.3: 49794706-49794706 on Build GRCh38
rs756974794 Chr.3: 49794646-49794646 on Build GRCh38
rs748612742 Chr.3: 49794693-49794693 on Build GRCh38

View Full Product Details